Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 000468
          Assay Name: hsa-miR-146a
          miRBase Accession Number: MI0000477
          miRBase Version: v22.1
          Chromosome Location: Chr.5: 160485352 - 160485450 [+] on Build GRCh38
          Mature miRNA Sequence: UGAGAACUGAAUUCCAUGGGUU
          Species: Human, Mouse, Rat, Callithrix jacchus, Chicken, Cricetulus griseus, Dog, Horse, Microcebus murinus, Monodelphis domestica, Nomascus leucogenys, Pan troglodytes, Papio hamadryas, Pig, Pongo pygmaeus, Pteropus alecto, Rhesus monkey, Taeniopygia guttata, Tupaia chinensis
          Product Type: TaqMan™ MicroRNA Assay
          Assay ID 000468
          Size
          Availability Inventoried
          Catalog # 4427975
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000103, mir-146

          Mature miRNA Sequence:

          UGAGAACUGAAUUCCAUGGGUU

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-146a-5p MIMAT0000449
          Mouse mmu-miR-146a-5p MIMAT0000158
          Rat rno-miR-146a-5p MIMAT0000852
          Callithrix jacchus cja-miR-146a MIMAT0039478
          Chicken gga-miR-146a-5p MIMAT0001163
          Cricetulus griseus cgr-miR-146a MIMAT0023774
          View More Rows
          Dog cfa-miR-146a MIMAT0006684
          Horse eca-miR-146a MIMAT0013065
          Microcebus murinus mmr-miR-146a MIMAT0049129
          Monodelphis domestica mdo-miR-146a-5p MIMAT0012746
          Nomascus leucogenys nle-miR-146a MIMAT0049418
          Pan troglodytes ptr-miR-146a MIMAT0008038
          Papio hamadryas pha-miR-146a MIMAT0049573
          Pig ssc-miR-146a-5p MIMAT0022963
          Pongo pygmaeus ppy-miR-146a MIMAT0015767
          Pteropus alecto pal-miR-146a-5p MIMAT0040181
          Rhesus monkey mml-miR-146a-5p MIMAT0006203
          Taeniopygia guttata tgu-miR-146c MIMAT0014609
          Tupaia chinensis tch-miR-146a-5p MIMAT0036567

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-146a
          Stem-loop Accession # MI0000477
          Stem-loop Sequence
          CCGAUGUGUAUCCUCAGCUUUGAGAACUGAAUUCCAUGGGUUGUGUCAGUGUCAGACCUCUGAAAUUCAGUUCUUCAGCUGGGAUAUCUCUGUCAUCGU
          Chromosome Location Chr. 5 - 160485352 - 160485450 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000449
          miRBase ID: hsa-miR-146a-5p
          Mature miRNA Sequence: UGAGAACUGAAUUCCAUGGGUU
          Chromosome Location: Chr. 5 - 160485352 - 160485450 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-146a
          Stem-loop Accession # MI0000170
          Stem-loop Sequence
          AGCUCUGAGAACUGAAUUCCAUGGGUUAUAUCAAUGUCAGACCUGUGAAAUUCAGUUCUUCAGCU
          Chromosome Location Chr. 11 - 43374397 - 43374461 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000158
          miRBase ID: mmu-miR-146a-5p
          Mature miRNA Sequence: UGAGAACUGAAUUCCAUGGGUU
          Chromosome Location: Chr. 11 - 43374397 - 43374461 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-146a
          Stem-loop Accession # MI0000919
          Stem-loop Sequence
          UGUGUAUCCUCAGCUCUGAGAACUGAAUUCCAUGGGUUAUAGCAAUGUCAGACCUGUGAAGUUCAGUUCUUUAGCUGGGAUAGCUCUAUCGUCAU
          Chromosome Location Chr. 10 - 28962476 - 28962570 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000852
          miRBase ID: rno-miR-146a-5p
          Mature miRNA Sequence: UGAGAACUGAAUUCCAUGGGUU
          Chromosome Location: Chr. 10 - 28962476 - 28962570 [-] on Build Rnor_6.0

          Callithrix jacchus

          Stem-loop ID cja-mir-146a
          Stem-loop Accession # MI0032003
          Stem-loop Sequence
          GUAUCCUCAGCUUUGAGAACUGAAUUCCAUGGGUUGUGUCAGUGUCAGACCUGUGAAAUUCAGUUCUUCAGCUGGGAUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039478
          miRBase ID: cja-miR-146a
          Mature miRNA Sequence: UGAGAACUGAAUUCCAUGGGUU
          Chromosome Location: NA

          Chicken

          Stem-loop ID gga-mir-146a
          Stem-loop Accession # MI0001235
          Stem-loop Sequence
          GUGUAUCCUCAGCUUUGAGAACUGAAUUCCAUGGGUUGUAAUUGAAUCCUUUGUCAGACCCAUGGGGCUCAGUUCUUCAGCUUGGAUAUUUCUGUCUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001163
          miRBase ID: gga-miR-146a-5p
          Mature miRNA Sequence: UGAGAACUGAAUUCCAUGGGUU
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-146a
          Stem-loop Accession # MI0020407
          Stem-loop Sequence
          CCUCAGCUCUGAGAACUGAAUUCCAUGGGUUGUGUCAAUGUCAGACCUGUGAAGUUCAGUUCUUCAGCUGGGAUAGCUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023774
          miRBase ID: cgr-miR-146a
          Mature miRNA Sequence: UGAGAACUGAAUUCCAUGGGUU
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-146a
          Stem-loop Accession # MI0008094
          Stem-loop Sequence
          UGAGAACUGAAUUCCAUGGGUUGUGUCAGUGUCAGACCUGUGAAGUUCAGUUCUUCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006684
          miRBase ID: cfa-miR-146a
          Mature miRNA Sequence: UGAGAACUGAAUUCCAUGGGUU
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-146a
          Stem-loop Accession # MI0012809
          Stem-loop Sequence
          AGCUUUGAGAACUGAAUUCCAUGGGUUGUGUCAGCGUCAGACCUGUGAAGUUCAGUUCUUCAGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013065
          miRBase ID: eca-miR-146a
          Mature miRNA Sequence: UGAGAACUGAAUUCCAUGGGUU
          Chromosome Location: NA

          Microcebus murinus

          Stem-loop ID mmr-mir-146a
          Stem-loop Accession # MI0039841
          Stem-loop Sequence
          UGAGAACUGAAUUCCAUGGGUUGUGUCAGUGUCAGACCUGUGAAUUUCAGUUCUUCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049129
          miRBase ID: mmr-miR-146a
          Mature miRNA Sequence: UGAGAACUGAAUUCCAUGGGUU
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-146a
          Stem-loop Accession # MI0012502
          Stem-loop Sequence
          UGUAUCUUCAGCUUUGAGAACUGAAUUCCAUGGGUUGUCUUUGUAUCAGACCUAUGAAACUCAGUUCUUCAGCUGGAAUAUCUCUGUCAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012746
          miRBase ID: mdo-miR-146a-5p
          Mature miRNA Sequence: UGAGAACUGAAUUCCAUGGGUU
          Chromosome Location: NA

          Nomascus leucogenys

          Stem-loop ID nle-mir-146a
          Stem-loop Accession # MI0040159
          Stem-loop Sequence
          UGAGAACUGAAUUCCAUGGGUUGUGUCAGUGUCAGACCUGUGAAAUUCAGUUCUUCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049418
          miRBase ID: nle-miR-146a
          Mature miRNA Sequence: UGAGAACUGAAUUCCAUGGGUU
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-146a
          Stem-loop Accession # MI0008544
          Stem-loop Sequence
          CGAUGUGUAUCCUCAGCUUUGAGAACUGAAUUCCAUGGGUUGUGUCAGUGUCAGACCUGUGAAAUUCAGUUCUUCAGCUGGGAUAUCUCUGUCAUCGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008038
          miRBase ID: ptr-miR-146a
          Mature miRNA Sequence: UGAGAACUGAAUUCCAUGGGUU
          Chromosome Location: NA

          Papio hamadryas

          Stem-loop ID pha-mir-146a
          Stem-loop Accession # MI0040344
          Stem-loop Sequence
          UGAGAACUGAAUUCCAUGGGUUGUGUCAGUGUCAGACCUGUGAAAUUCAGUUCUUCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049573
          miRBase ID: pha-miR-146a
          Mature miRNA Sequence: UGAGAACUGAAUUCCAUGGGUU
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-146a
          Stem-loop Accession # MI0015926
          Stem-loop Sequence
          UACUCAGCUUUGAGAACUGAAUUCCAUGGGUUGUGUCAUUGUCAGACCUGUGAAGUUUAGUUCUUCAGCUGGGAUAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0022963
          miRBase ID: ssc-miR-146a-5p
          Mature miRNA Sequence: UGAGAACUGAAUUCCAUGGGUU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-146a
          Stem-loop Accession # MI0014834
          Stem-loop Sequence
          CCGAUGUGUAUCCUCAGCUUUGAGAACUGAAUUCCAUGGGUUGUGUCUGUGUCAGACCUGUGAAAUUCAGUUCUUCAGCUGGGAUAUCUCUGUCGUCGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015767
          miRBase ID: ppy-miR-146a
          Mature miRNA Sequence: UGAGAACUGAAUUCCAUGGGUU
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-146a
          Stem-loop Accession # MI0032537
          Stem-loop Sequence
          UGAGAACUGAAUUCCAUGGGUUGUGUCAGUGUCAGACCUGUGAAAUUCAGUUCUUCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0040181
          miRBase ID: pal-miR-146a-5p
          Mature miRNA Sequence: UGAGAACUGAAUUCCAUGGGUU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-146a
          Stem-loop Accession # MI0007634
          Stem-loop Sequence
          CCUAUGUGUAUCCUCAGCUUUGAGAACUGAAUUCCAUGGGUUGUGUCAGUGUCAGACCUGUGAAAUUCAGUUCUUCAGCUGGGAUAUCUCUGUCGUCGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006203
          miRBase ID: mml-miR-146a-5p
          Mature miRNA Sequence: UGAGAACUGAAUUCCAUGGGUU
          Chromosome Location: NA

          Taeniopygia guttata

          Stem-loop ID tgu-mir-146c
          Stem-loop Accession # MI0013839
          Stem-loop Sequence
          UUCUCAGCUUUGAGAACUGAAUUCCAUGGGUUGUAAUUGAAUCUGCUGUCAGACCCAUGGGGCUCAGUUCUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014609
          miRBase ID: tgu-miR-146c
          Mature miRNA Sequence: UGAGAACUGAAUUCCAUGGGUU
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-mir-146a
          Stem-loop Accession # MI0031230
          Stem-loop Sequence
          UGAGAACUGAAUUCCAUGGGUUGUGUCAGCGUCAGACCUGUGAAAUUCAGUUCUUCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036567
          miRBase ID: tch-miR-146a-5p
          Mature miRNA Sequence: UGAGAACUGAAUUCCAUGGGUU
          Chromosome Location: NA


          Back To Top

          More Information


          Associated Megaplex™ RT Primer Pools:

          Megaplex™ RT Primers, Human Pool A
          Megaplex™ RT Primers, Rodent Pool A




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03465077_pri
          TaqMan™ Pri-miRNA Assay : Mm03306349_pri
          TaqMan™ Pri-miRNA Assay : Hs03303259_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10722
          Ambion® Pre-miR™ miRNA Precursor : PM10722
          mirVana® miRNA inhibitor : MH10722
          mirVana® miRNA mimic : MC10722
          TaqMan™ Advanced miRNA Assay : mmu478399_mir
          TaqMan™ Advanced miRNA Assay : 478399_mir
          TaqMan™ Advanced miRNA Assay : rno481451_mir
          MIRNA : TaqMan® OpenArray Rodent MicroRNA Panel
          FGMIR : TaqManTM Human MicroRNA Array A
          miRNA Pool : Megaplex™ RT Primers, Human Pool A
          FGMIR : TaqManTM Rodent MicroRNA Array A
          MIRNA : TaqMan® Array Rodent MicroRNA A+B Cards Set v3.0
          MIRNA : TaqMan® Array Rodent MicroRNA A Card v2.0
          FGMIR : QS TaqMan OpenArray Hu miRNA Plate
          miRNA Card : TaqMan® Human MicroRNA A Cards v2.0
          miRNA Card : TaqMan® Rodent MicroRNA A Cards v2.0
          FGMIR : QS TaqMan OpenArray Rod miRNA Plate
          MIRNA : TaqMan® OpenArray Human MicroRNA Panel
          MIRNA : TaqMan® Array Human MicroRNA A Cards v2.0
          MIRNA : TaqMan® Array Human MicroRNA A+B Cards Set v3.0
          miRNA Pool : Megaplex™ RT Primers, Rodent Pool A


          miRBase Alias:

          Human: hsa-miR-146a (v17)
          Mouse: mmu-miR-146a (v17)
          Mouse: mmu-miR-146 (v9.2)
          Rat: rno-miR-146a (v18)
          Rat: rno-miR-146 (v9.2)
          Gallus gallus: gga-miR-146 (v7.1)
          Gallus gallus: gga-miR-146a (v19)
          Macaca mulatta: mml-miR-146a (v19)
          Monodelphis domestica: mdo-miR-146a (v19)

          Back To Top

          Related Products

          • TaqMan® Universal Master Mix II, with UNG
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® MicroRNA Reverse Transcription Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
          git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.49.0-Offline