Search
Search

| Assay Name: | snoRNA234 |
|---|---|
| NCBI Accession Number: | AF357329 |
| Control Sequence: | CTTTTGGAACTGAATCTAAGTGATTTAACAAAAATTCGTCACTACCACTGAGA |
| Species: | Mouse |
| Product Type: | TaqMan™ microRNA Control Assay |
Alias: | snoRNA234, snoRNA |
Control Sequence: | CTTTTGGAACTGAATCTAAGTGATTTAACAAAAATTCGTCACTACCACTGAGA |
| Species | NCBI Accession Number |
|---|---|
| AF357329 |