Search
Search

| Assay Name: | hsa-miR-455-3p |
|---|---|
| miRBase Accession Number: | MI0003513 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GCAGUCCAUGGGCAUAUACAC |
| Species: | Human, Anolis carolinensis, Gorilla gorilla, Ophiophagus hannah, Pan troglodytes, Pig, Pongo pygmaeus, Python bivittatus, Rhesus monkey |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000129, mir-455 |
Mature miRNA Sequence: | GCAGUCCAUGGGCAUAUACAC |
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-455-3p | MIMAT0004784 | |
| Anolis carolinensis | aca-miR-455-3p | MIMAT0021965 | |
| Gorilla gorilla | ggo-miR-455 | MIMAT0024222 | |
| Ophiophagus hannah | oha-miR-455-3p | MIMAT0036932 | |
| Pan troglodytes | ptr-miR-455 | MIMAT0008157 | |
| Pig | ssc-miR-455-3p | MIMAT0013960 | |
| View More Rows | |||
| Pongo pygmaeus | ppy-miR-455-3p | MIMAT0015900 | |
| Python bivittatus | pbv-miR-455-3p | MIMAT0039058 | |
| Rhesus monkey | mml-miR-455-3p | MIMAT0006340 | |