Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 002253
          Assay Name: hsa-miR-101
          miRBase Accession Number: MI0000103
          miRBase Version: v22.1
          Chromosome Location: Chr.1: 65058434 - 65058508 [-] on Build GRCh38
          Mature miRNA Sequence: UACAGUACUGUGAUAACUGAA
          Species: Human, Mouse, Rat, Bovine, Callithrix jacchus, Columba livia, Horse, Pig, Pongo pygmaeus
          Product Type: TaqMan™ MicroRNA Assay
          Assay ID 002253
          Size
          Availability Inventoried
          Catalog # 4427975
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000046, mir-101

          Mature miRNA Sequence:

          UACAGUACUGUGAUAACUGAA

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-101-3p MIMAT0000099
          Mouse mmu-miR-101a-3p MIMAT0000133
          Rat rno-miR-101a-3p MIMAT0000823
          Bovine bta-miR-101 MIMAT0003520
          Callithrix jacchus cja-miR-101-2 MIMAT0049331
          Columba livia cli-miR-101-3p MIMAT0038483
          View More Rows
          Horse eca-miR-101 MIMAT0012951
          Pig ssc-miR-101 MIMAT0010185
          Pongo pygmaeus ppy-miR-101 MIMAT0002429

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-101-1
          Stem-loop Accession # MI0000103
          Stem-loop Sequence
          UGCCCUGGCUCAGUUAUCACAGUGCUGAUGCUGUCUAUUCUAAAGGUACAGUACUGUGAUAACUGAAGGAUGGCA
          Chromosome Location Chr. 1 - 65058434 - 65058508 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000099
          miRBase ID: hsa-miR-101-3p
          Mature miRNA Sequence: UACAGUACUGUGAUAACUGAA
          Chromosome Location: Chr. 1 - 65058434 - 65058508 [-] on Build GRCh38

          Stem-loop ID hsa-mir-101-2
          Stem-loop Accession # MI0000739
          Stem-loop Sequence
          ACUGUCCUUUUUCGGUUAUCAUGGUACCGAUGCUGUAUAUCUGAAAGGUACAGUACUGUGAUAACUGAAGAAUGGUGGU
          Chromosome Location Chr. 9 - 4850297 - 4850375 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000099
          miRBase ID: hsa-miR-101-3p
          Mature miRNA Sequence: UACAGUACUGUGAUAACUGAA
          Chromosome Location: Chr. 1 - 65058434 - 65058508 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-101a
          Stem-loop Accession # MI0000148
          Stem-loop Sequence
          AGGCUGCCCUGGCUCAGUUAUCACAGUGCUGAUGCUGUCCAUUCUAAAGGUACAGUACUGUGAUAACUGAAGGAUGGCAGCCA
          Chromosome Location Chr. 4 - 101346945 - 101347027 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000133
          miRBase ID: mmu-miR-101a-3p
          Mature miRNA Sequence: UACAGUACUGUGAUAACUGAA
          Chromosome Location: Chr. 4 - 101346945 - 101347027 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-101a
          Stem-loop Accession # MI0000886
          Stem-loop Sequence
          UGCCCUGGCUCAGUUAUCACAGUGCUGAUGCUGUCCAUUCUAAAGGUACAGUACUGUGAUAACUGAAGGAUGGCA
          Chromosome Location Chr. 5 - 120168437 - 120168511 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000823
          miRBase ID: rno-miR-101a-3p
          Mature miRNA Sequence: UACAGUACUGUGAUAACUGAA
          Chromosome Location: Chr. 5 - 120168437 - 120168511 [-] on Build Rnor_6.0

          Bovine

          Stem-loop ID bta-mir-101-1
          Stem-loop Accession # MI0009721
          Stem-loop Sequence
          AGGCUGCCCUGGCUCAGUUAUCACAGUGCUGAUGCUGUCCAUUCUAAAGGUACAGUACUGUGAUAACUGAAGGAUGGCAGCCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003520
          miRBase ID: bta-miR-101
          Mature miRNA Sequence: UACAGUACUGUGAUAACUGAA
          Chromosome Location: NA

          Stem-loop ID bta-mir-101-2
          Stem-loop Accession # MI0004735
          Stem-loop Sequence
          ACUGUCCUUUUUCGGUUAUCAUGGUACCGAUGCUGUAUAUCUGAAAGGUACAGUACUGUGAUAACUGAAGAAUGGUGGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003520
          miRBase ID: bta-miR-101
          Mature miRNA Sequence: UACAGUACUGUGAUAACUGAA
          Chromosome Location: NA

          Callithrix jacchus

          Stem-loop ID cja-mir-101-2
          Stem-loop Accession # MI0040067
          Stem-loop Sequence
          CGGUUAUCAUGGUACCGAUGCUGUAUAUCUGAAAGGUACAGUACUGUGAUAACUGAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049331
          miRBase ID: cja-miR-101-2
          Mature miRNA Sequence: UACAGUACUGUGAUAACUGAA
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-mir-101-1
          Stem-loop Accession # MI0029947
          Stem-loop Sequence
          ACAGGCUGCCCUGGCUCAGUUAUCACAGUGCUGAUGCUGUCUCUUGUAAAGGUACAGUACUGUGAUAACUGAAGGAUGGCAGCCAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038483
          miRBase ID: cli-miR-101-3p
          Mature miRNA Sequence: UACAGUACUGUGAUAACUGAA
          Chromosome Location: NA

          Stem-loop ID cli-mir-101-2
          Stem-loop Accession # MI0029948
          Stem-loop Sequence
          ACUAUGAACUGUCCUUUUUCGGUUAUCAUGGUACCGGUGCUGUAUACGUGAAAGGUACAGUACUGUGAUAACUGAAGAAUGGUGGUGCCAUCAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038483
          miRBase ID: cli-miR-101-3p
          Mature miRNA Sequence: UACAGUACUGUGAUAACUGAA
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-101-1
          Stem-loop Accession # MI0012703
          Stem-loop Sequence
          CUCAGUUAUCACAGUGCUGAUGCUGUCAGUUCUAAAGGUACAGUACUGUGAUAACUGAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012951
          miRBase ID: eca-miR-101
          Mature miRNA Sequence: UACAGUACUGUGAUAACUGAA
          Chromosome Location: NA

          Stem-loop ID eca-mir-101-2
          Stem-loop Accession # MI0012861
          Stem-loop Sequence
          CCUUUUUCGGUUAUCAUGGUACCGAUGCUGUAUAUCUGAAAGGUACAGUACUGUGAUAACUGAAGAAUGGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012951
          miRBase ID: eca-miR-101
          Mature miRNA Sequence: UACAGUACUGUGAUAACUGAA
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-101-1
          Stem-loop Accession # MI0010678
          Stem-loop Sequence
          GACUGAACUGUCCUUUUUCGGUUAUCAUGGUACCGAUGCUGUAUAUCUGAAAGGUACAGUACUGUGAUAACUGAAGAAUGGUGGUGCCAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0010185
          miRBase ID: ssc-miR-101
          Mature miRNA Sequence: UACAGUACUGUGAUAACUGAA
          Chromosome Location: NA

          Stem-loop ID ssc-mir-101-2
          Stem-loop Accession # MI0010679
          Stem-loop Sequence
          GGCUGCCCUGGCUCAGUUAUCACAGUGCUGAUGCUGUCCAUUCUAAAGGUACAGUACUGUGAUAACUGAAGGAUGGCAGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0010185
          miRBase ID: ssc-miR-101
          Mature miRNA Sequence: UACAGUACUGUGAUAACUGAA
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-101-1
          Stem-loop Accession # MI0014811
          Stem-loop Sequence
          UGCCCUGGCUCAGUUAUCACAGUGCUGAUGCUGUCCAUUCUAAAGGUACAGUACUGUGAUAACUGAAGGAUGGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002429
          miRBase ID: ppy-miR-101
          Mature miRNA Sequence: UACAGUACUGUGAUAACUGAA
          Chromosome Location: NA

          Stem-loop ID ppy-mir-101-2
          Stem-loop Accession # MI0014812
          Stem-loop Sequence
          ACUGUCCUUUUUCGGUUAUCAUGGUACCGAUGCUGUAUAUCUGAAAGGUACAGUACUGUGAUAACUGAAGAAUGGUGGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002429
          miRBase ID: ppy-miR-101
          Mature miRNA Sequence: UACAGUACUGUGAUAACUGAA
          Chromosome Location: NA


          Back To Top

          More Information


          Associated Megaplex™ RT Primer Pools:

          Megaplex™ RT Primers, Human Pool A
          Megaplex™ RT Primers, Rodent Pool A




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03306215_pri
          TaqMan™ Pri-miRNA Assay : Rn03464794_pri
          TaqMan™ Pri-miRNA Assay : Hs03302741_pri
          TaqMan™ Pri-miRNA Assay : Hs03303387_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM11414
          Ambion® Pre-miR™ miRNA Precursor : PM11414
          mirVana® miRNA inhibitor : MH11414
          mirVana® miRNA mimic : MC11414
          TaqMan™ Advanced miRNA Assay : 477863_mir
          TaqMan™ Advanced miRNA Assay : mmu483018_mir
          MIRNA : TaqMan® Array Human MicroRNA A+B Cards Set v3.0
          miRNA Card : TaqMan® Human MicroRNA A Cards v2.0
          MIRNA : TaqMan® Array Human MicroRNA A Cards v2.0
          FGMIR : QS TaqMan OpenArray Hu miRNA Plate
          miRNA Pool : Megaplex™ RT Primers, Human Pool A
          MIRNA : TaqMan® OpenArray Rodent MicroRNA Panel
          FGMIR : QS TaqMan OpenArray Rod miRNA Plate
          MIRNA : TaqMan® Array Rodent MicroRNA A+B Cards Set v3.0
          miRNA Card : TaqMan® Rodent MicroRNA A Cards v2.0
          miRNA Pool : Megaplex™ RT Primers, Rodent Pool A
          FGMIR : TaqManTM Rodent MicroRNA Array A
          FGMIR : TaqManTM Human MicroRNA Array A
          MIRNA : TaqMan® OpenArray Human MicroRNA Panel
          MIRNA : TaqMan® Array Rodent MicroRNA A Card v2.0


          miRBase Alias:

          Human: hsa-miR-101 (v17)
          Mouse: mmu-miR-101a (v17)
          Rat: rno-miR-101a (v18)
          Staphylococcus schleiferi: ssc-miR-101a (v14)

          Back To Top

          Related Products

          • TaqMan® Universal Master Mix II, with UNG
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® MicroRNA Reverse Transcription Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline