Search
Search

| Assay Name: | hsa-miR-422a |
|---|---|
| miRBase Accession Number: | MI0001444 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | ACUGGACUUAGGGUCAGAAGGC |
| Species: | Human, Pan troglodytes |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000548, mir-422 |
Mature miRNA Sequence: | ACUGGACUUAGGGUCAGAAGGC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-422a | MIMAT0001339 |
| Pan troglodytes | ptr-miR-422a | MIMAT0008141 |