Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 002324
          Assay Name: hsa-miR-744
          miRBase Accession Number: MI0005559
          miRBase Version: v22.1
          Chromosome Location: Chr.17: 12081899 - 12081996 [+] on Build GRCh38
          Mature miRNA Sequence: UGCGGGGCUAGGGCUAACAGCA
          Species: Human, Mouse, Bovine, Dasypus novemcinctus, Guinea pig, Pan troglodytes, Pig, Pongo pygmaeus, Rabbit
          Product Type: TaqMan™ MicroRNA Assay
          Assay ID 002324
          Size
          Availability Inventoried
          Catalog # 4427975
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000431, mir-744

          Mature miRNA Sequence:

          UGCGGGGCUAGGGCUAACAGCA

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-744-5p MIMAT0004945
          Mouse mmu-miR-744-5p MIMAT0004187
          Bovine bta-miR-744 MIMAT0009369
          Dasypus novemcinctus dno-miR-744-5p MIMAT0048008
          Guinea pig cpo-miR-744-5p MIMAT0047379
          Pan troglodytes ptr-miR-744 MIMAT0008317
          View More Rows
          Pig ssc-miR-744 MIMAT0015708
          Pongo pygmaeus ppy-miR-744 MIMAT0016102
          Rabbit ocu-miR-744-5p MIMAT0048563

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-744
          Stem-loop Accession # MI0005559
          Stem-loop Sequence
          UUGGGCAAGGUGCGGGGCUAGGGCUAACAGCAGUCUUACUGAAGGUUUCCUGGAAACCACGCACAUGCUGUUGCCACUAACCUCAACCUUACUCGGUC
          Chromosome Location Chr. 17 - 12081899 - 12081996 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004945
          miRBase ID: hsa-miR-744-5p
          Mature miRNA Sequence: UGCGGGGCUAGGGCUAACAGCA
          Chromosome Location: Chr. 17 - 12081899 - 12081996 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-744
          Stem-loop Accession # MI0004124
          Stem-loop Sequence
          GGCUGGGCAAGGUGCGGGGCUAGGGCUAACAGCAGUCUUACUGACGGUUUCCUGGAAACCACACACAUGCUGUUGCCACUAACCUCAACCUUACUCGGUC
          Chromosome Location Chr. 11 - 65734733 - 65734832 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004187
          miRBase ID: mmu-miR-744-5p
          Mature miRNA Sequence: UGCGGGGCUAGGGCUAACAGCA
          Chromosome Location: Chr. 11 - 65734733 - 65734832 [-] on Build GRCm38

          Bovine

          Stem-loop ID bta-mir-744
          Stem-loop Accession # MI0009891
          Stem-loop Sequence
          GGUUGGGCGGGGUGCGGGGCUAGGGCUAACAGCAGGCUCACUGACGGUUUCCCGGAAACCACGCACAUGCUGUUGCCACUAACCUCAACCUUACUCGGUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009369
          miRBase ID: bta-miR-744
          Mature miRNA Sequence: UGCGGGGCUAGGGCUAACAGCA
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-744
          Stem-loop Accession # MI0039171
          Stem-loop Sequence
          UGCGGGGCUAGGGCUAACAGCAGUCUUACUGAAGGUUUCCUGGAAACCACACACAUGCUGUUGCCACUAACCUCAACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048008
          miRBase ID: dno-miR-744-5p
          Mature miRNA Sequence: UGCGGGGCUAGGGCUAACAGCA
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-744
          Stem-loop Accession # MI0038839
          Stem-loop Sequence
          UGCGGGGCUAGGGCUAACAGCAGUCUUACUGAAGGUUUCCUGGAAACCACACACAUGCUGUUGCCACUAACCUCAACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047379
          miRBase ID: cpo-miR-744-5p
          Mature miRNA Sequence: UGCGGGGCUAGGGCUAACAGCA
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-744
          Stem-loop Accession # MI0008856
          Stem-loop Sequence
          UUGGGCAAGGUGCGGGGCUAGGGCUAACAGCAGUCUUACUGAAGGUUUCCUGGAAACCACGCACAUGCUGUUGCCACUAACCUCAACCUUACUCGGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008317
          miRBase ID: ptr-miR-744
          Mature miRNA Sequence: UGCGGGGCUAGGGCUAACAGCA
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-744
          Stem-loop Accession # MI0014769
          Stem-loop Sequence
          GCAGGGUGCGGGGCUAGGGCUAACAGCAGUCUUACUGAAGGUUUCCUGGAAACCACGCACAUGCUGUUGCCACUAACCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015708
          miRBase ID: ssc-miR-744
          Mature miRNA Sequence: UGCGGGGCUAGGGCUAACAGCA
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-744
          Stem-loop Accession # MI0015142
          Stem-loop Sequence
          UUGGGCAAGGUGCGGGGCUAGGGCUAACAGCAGUCUUACUGAAGGUUUCCUGGAAACCACGCACAUGCUGUUGCCACUAACCUCAACCUUACUCGGUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0016102
          miRBase ID: ppy-miR-744
          Mature miRNA Sequence: UGCGGGGCUAGGGCUAACAGCA
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-744
          Stem-loop Accession # MI0039464
          Stem-loop Sequence
          UGCGGGGCUAGGGCUAACAGCAGUCUUACUGAAGGUUUCCUGGAAACCACACACAUGCUGUUGCCACUAACCUCAACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048563
          miRBase ID: ocu-miR-744-5p
          Mature miRNA Sequence: UGCGGGGCUAGGGCUAACAGCA
          Chromosome Location: NA


          Back To Top

          More Information


          Associated Megaplex™ RT Primer Pools:

          Megaplex™ RT Primers, Human Pool A
          Megaplex™ RT Primers, Rodent Pool A




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03307769_pri
          TaqMan™ Pri-miRNA Assay : Hs03294362_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM13027
          Ambion® Pre-miR™ miRNA Precursor : PM13027
          mirVana® miRNA inhibitor : MH13027
          mirVana® miRNA mimic : MC13027
          TaqMan™ Advanced miRNA Assay : 478200_mir
          MIRNA : TaqMan® Array Rodent MicroRNA A+B Cards Set v3.0
          MIRNA : TaqMan® Array Human MicroRNA A Cards v2.0
          MIRNA : TaqMan® Array Human MicroRNA A+B Cards Set v3.0
          MIRNA : TaqMan® OpenArray Human MicroRNA Panel
          MIRNA : TaqMan® OpenArray Rodent MicroRNA Panel
          FGMIR : QS TaqMan OpenArray Hu miRNA Plate
          MIRNA : TaqMan® Array Rodent MicroRNA A Card v2.0
          FGMIR : TaqManTM Human MicroRNA Array A
          miRNA Pool : Megaplex™ RT Primers, Human Pool A
          miRNA Card : TaqMan® Human MicroRNA A Cards v2.0
          FGMIR : QS TaqMan OpenArray Rod miRNA Plate
          FGMIR : TaqManTM Rodent MicroRNA Array A
          miRNA Pool : Megaplex™ RT Primers, Rodent Pool A
          miRNA Card : TaqMan® Rodent MicroRNA A Cards v2.0


          miRBase Alias:

          Human: hsa-miR-744 (v17)
          Mouse: mmu-miR-744 (v17)

          Back To Top

          Related Products

          • TaqMan® Universal Master Mix II, with UNG
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® MicroRNA Reverse Transcription Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline