Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 002623
          Assay Name: hsa-miR-155
          miRBase Accession Number: MI0033871
          miRBase Version: v22.1
          Chromosome Location: -
          Mature miRNA Sequence: UUAAUGCUAAUCGUGAUAGGGGU
          Species: Astatotilapia burtoni, Bovine, Columba livia, Dog, Gadus morhua, Goat, Horse, Metriaclima zebra, Neolamprologus brichardi, Oreochromis niloticus, Ornithorhynchus anatinus, Pan troglodytes, Papio hamadryas, Pongo pygmaeus, Pteropus alecto, Pundamilia nyererei, Rhesus monkey, Salmo salar, Tupaia chinensis
          Product Type: TaqMan™ MicroRNA Assay
          Assay ID 002623
          Size
          Availability Inventoried
          Catalog # 4427975
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Mature miRNA Sequence:

          UUAAUGCUAAUCGUGAUAGGGGU

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Astatotilapia burtoni abu-miR-155 MIMAT0042155
          Bovine bta-miR-155 MIMAT0009241
          Columba livia cli-miR-155-5p MIMAT0038549
          Dog cfa-miR-155 MIMAT0006671
          Gadus morhua gmo-miR-155-5p MIMAT0044060
          Goat chi-miR-155-5p MIMAT0035988
          View More Rows
          Horse eca-miR-155 MIMAT0013182
          Metriaclima zebra mze-miR-155 MIMAT0042397
          Neolamprologus brichardi nbr-miR-155 MIMAT0042571
          Oreochromis niloticus oni-miR-155 MIMAT0042751
          Ornithorhynchus anatinus oan-miR-155-5p MIMAT0006988
          Pan troglodytes ptr-miR-155 MIMAT0008048
          Papio hamadryas pha-miR-155 MIMAT0049558
          Pongo pygmaeus ppy-miR-155 MIMAT0015777
          Pteropus alecto pal-miR-155-5p MIMAT0039975
          Pundamilia nyererei pny-miR-155 MIMAT0042961
          Rhesus monkey mml-miR-155 MIMAT0006216
          Salmo salar ssa-miR-155-5p MIMAT0032392
          Tupaia chinensis tch-miR-155-5p MIMAT0036472

          Stem-loop Details



          Astatotilapia burtoni

          Stem-loop ID abu-mir-155
          Stem-loop Accession # MI0033871
          Stem-loop Sequence
          UUAAUGCUAAUCGUGAUAGGGGUUGUUCUUAUUAACAGACACCUAACAUGUUAGCAUUAGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042155
          miRBase ID: abu-miR-155
          Mature miRNA Sequence: UUAAUGCUAAUCGUGAUAGGGGU
          Chromosome Location: NA

          Bovine

          Stem-loop ID bta-mir-155
          Stem-loop Accession # MI0009752
          Stem-loop Sequence
          UGUUAAUGCUAAUCGUGAUAGGGGUUUUUACCUCGGACUGACUCCUACAUGUUAGCAUUAACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009241
          miRBase ID: bta-miR-155
          Mature miRNA Sequence: UUAAUGCUAAUCGUGAUAGGGGU
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-mir-155
          Stem-loop Accession # MI0029988
          Stem-loop Sequence
          UGUAGGCUGUAUGUUGUUAAUGCUAAUCGUGAUAGGGGUUUUUACCUCUGACUGACUCCUACAUAUUAGCAUUAACACUGUAUCAUGCCUCUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038549
          miRBase ID: cli-miR-155-5p
          Mature miRNA Sequence: UUAAUGCUAAUCGUGAUAGGGGU
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-155
          Stem-loop Accession # MI0008078
          Stem-loop Sequence
          UUAAUGCUAAUCGUGAUAGGGGUUUUUACCUCCGACUGACUCCUACAUGUUAGCAUUAACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006671
          miRBase ID: cfa-miR-155
          Mature miRNA Sequence: UUAAUGCUAAUCGUGAUAGGGGU
          Chromosome Location: NA

          Gadus morhua

          Stem-loop ID gmo-mir-155
          Stem-loop Accession # MI0036008
          Stem-loop Sequence
          UUAAUGCUAAUCGUGAUAGGGGUUGUUCUCUCCAAGACGCCUAGCAUGCUAGCAUUAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0044060
          miRBase ID: gmo-miR-155-5p
          Mature miRNA Sequence: UUAAUGCUAAUCGUGAUAGGGGU
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-155
          Stem-loop Accession # MI0030643
          Stem-loop Sequence
          UUGCUGUGGGCUGUGUGCUAUUAAUGCUAAUCGUGAUAGGGGUUUUUACCUCGGACUGACUCCUACAUGUUAGCAUUAACAGGGCAUGGCGCCUGUUAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0035988
          miRBase ID: chi-miR-155-5p
          Mature miRNA Sequence: UUAAUGCUAAUCGUGAUAGGGGU
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-155
          Stem-loop Accession # MI0012927
          Stem-loop Sequence
          CUGUUAAUGCUAAUCGUGAUAGGGGUUUUUACCUCGGACUGACUCCUACAUGUUAGCAUUAACAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013182
          miRBase ID: eca-miR-155
          Mature miRNA Sequence: UUAAUGCUAAUCGUGAUAGGGGU
          Chromosome Location: NA

          Metriaclima zebra

          Stem-loop ID mze-mir-155
          Stem-loop Accession # MI0034188
          Stem-loop Sequence
          UUAAUGCUAAUCGUGAUAGGGGUUGUUCUUAUUAACAGACACCUAACAUGUUAGCAUUAGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042397
          miRBase ID: mze-miR-155
          Mature miRNA Sequence: UUAAUGCUAAUCGUGAUAGGGGU
          Chromosome Location: NA

          Neolamprologus brichardi

          Stem-loop ID nbr-mir-155
          Stem-loop Accession # MI0034429
          Stem-loop Sequence
          UUAAUGCUAAUCGUGAUAGGGGUUGUUCUUAUUAACAGACACCUAACAUGUUAGCAUUAGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042571
          miRBase ID: nbr-miR-155
          Mature miRNA Sequence: UUAAUGCUAAUCGUGAUAGGGGU
          Chromosome Location: NA

          Oreochromis niloticus

          Stem-loop ID oni-mir-155
          Stem-loop Accession # MI0034677
          Stem-loop Sequence
          UUAAUGCUAAUCGUGAUAGGGGUUGUUCUUAUUAACAGACACCUAACAUGUUAGCAUUAGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042751
          miRBase ID: oni-miR-155
          Mature miRNA Sequence: UUAAUGCUAAUCGUGAUAGGGGU
          Chromosome Location: NA

          Ornithorhynchus anatinus

          Stem-loop ID oan-mir-155
          Stem-loop Accession # MI0006775
          Stem-loop Sequence
          GGGACGCCGUGGGCCGCAGGUCGUUAAUGCUAAUCGUGAUAGGGGUUUUCUCUGUCUGACUGACCCCUGCCCGUUAGCAUUAGCAACGUGCGGCCCCUCGAACCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006988
          miRBase ID: oan-miR-155-5p
          Mature miRNA Sequence: UUAAUGCUAAUCGUGAUAGGGGU
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-155
          Stem-loop Accession # MI0008554
          Stem-loop Sequence
          UGUUAAUGCUAAUCGUGAUAGGGGUUUUUGCCUCCAAAUGACUCCUACAUAUUAGCAUUAACAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008048
          miRBase ID: ptr-miR-155
          Mature miRNA Sequence: UUAAUGCUAAUCGUGAUAGGGGU
          Chromosome Location: NA

          Papio hamadryas

          Stem-loop ID pha-mir-155
          Stem-loop Accession # MI0040328
          Stem-loop Sequence
          UUAAUGCUAAUCGUGAUAGGGGUUUUUACCUCCAACUGACUCCUACAUGUUAGCAUUAACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049558
          miRBase ID: pha-miR-155
          Mature miRNA Sequence: UUAAUGCUAAUCGUGAUAGGGGU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-155
          Stem-loop Accession # MI0014843
          Stem-loop Sequence
          CUGUUAAUGCUAAUCGUGAUAGGGGUUUUUACCUCCAACUGACUCCUACAUGUUAGCAUUAACAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015777
          miRBase ID: ppy-miR-155
          Mature miRNA Sequence: UUAAUGCUAAUCGUGAUAGGGGU
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-155
          Stem-loop Accession # MI0032429
          Stem-loop Sequence
          UUAAUGCUAAUCGUGAUAGGGGUUUUACCUCCGACUGACUCCUACAUGUUAGCAUUAACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039975
          miRBase ID: pal-miR-155-5p
          Mature miRNA Sequence: UUAAUGCUAAUCGUGAUAGGGGU
          Chromosome Location: NA

          Pundamilia nyererei

          Stem-loop ID pny-mir-155
          Stem-loop Accession # MI0034951
          Stem-loop Sequence
          UUAAUGCUAAUCGUGAUAGGGGUUGUUCUUAUUAACAGACACCUAACAUGUUAGCAUUAGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042961
          miRBase ID: pny-miR-155
          Mature miRNA Sequence: UUAAUGCUAAUCGUGAUAGGGGU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-155
          Stem-loop Accession # MI0007645
          Stem-loop Sequence
          CUGUUAAUGCUAAUCGUGAUAGGGGUUUUUACCUCCAACUGACUCCUACAUGUUAGCAUUAACAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006216
          miRBase ID: mml-miR-155
          Mature miRNA Sequence: UUAAUGCUAAUCGUGAUAGGGGU
          Chromosome Location: NA

          Salmo salar

          Stem-loop ID ssa-mir-155-1
          Stem-loop Accession # MI0026520
          Stem-loop Sequence
          UUAAUGCUAAUCGUGAUAGGGGUUCUGUUGUUUAAAAAAAAGACACCUAGCAUGUAAGCAUUAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032392
          miRBase ID: ssa-miR-155-5p
          Mature miRNA Sequence: UUAAUGCUAAUCGUGAUAGGGGU
          Chromosome Location: NA

          Stem-loop ID ssa-mir-155-2
          Stem-loop Accession # MI0026521
          Stem-loop Sequence
          UUAAUGCUAAUCGUGAUAGGGGUUCUGUUGUUAAAAAAAAAGACACCUAGCAUGUAAGCAUUAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032392
          miRBase ID: ssa-miR-155-5p
          Mature miRNA Sequence: UUAAUGCUAAUCGUGAUAGGGGU
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-mir-155
          Stem-loop Accession # MI0031131
          Stem-loop Sequence
          UUAAUGCUAAUCGUGAUAGGGGUUUUUGCCUCCGACUGACUCCUACAUGUUAGCAUUAACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036472
          miRBase ID: tch-miR-155-5p
          Mature miRNA Sequence: UUAAUGCUAAUCGUGAUAGGGGU
          Chromosome Location: NA


          Back To Top

          More Information


          Associated Megaplex™ RT Primer Pool:

          Megaplex™ RT Primers, Human Pool A




          Related Products

          Ambion® Anti-miR™ miRNA Inhibitor : AM12601
          Ambion® Pre-miR™ miRNA Precursor : PM12601
          mirVana® miRNA inhibitor : MH12601
          mirVana® miRNA mimic : MC12601
          TaqMan™ Advanced miRNA Assay : 477927_mir
          FGMIR : TaqManTM Human MicroRNA Array A
          MIRNA : TaqMan® Array Human MicroRNA A Cards v2.0
          miRNA Card : TaqMan® Human MicroRNA A Cards v2.0
          FGMIR : QS TaqMan OpenArray Rod miRNA Plate
          MIRNA : TaqMan® OpenArray Rodent MicroRNA Panel
          MIRNA : TaqMan® Array Human MicroRNA A+B Cards Set v3.0
          miRNA Pool : Megaplex™ RT Primers, Human Pool A


          miRBase Alias:

          Human: hsa-miR-155 (v17)
          Human: hsa-miR-155-5p (v21)
          Ochrobactrum anthropi: oan-miR-155 (v18)

          Back To Top

          Related Products

          • TaqMan® Universal Master Mix II, with UNG
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® MicroRNA Reverse Transcription Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline