Search
Search

| Assay Name: | csi-miR167a |
|---|---|
| miRBase Accession Number: | MI0040573 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGAAGCUGCCAGCAUGAUCUG |
| Species: | Amborella trichopoda, Asparagus officinalis, Citrus sinensis, Cucumis melo, Digitalis purpurea, Glycine max, Glycine soja, Linum usitatissimum, Lotus japonicus, Manihot esculenta, Medicago truncatula, Populus trichocarpa, Rice, Saccharum officinarum, Saccharum sp., Sorghum bicolor, Vitis vinifera, Zea mays |
| Product Type: | TaqMan™ MicroRNA Assay |
Mature miRNA Sequence: | UGAAGCUGCCAGCAUGAUCUG |
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Amborella trichopoda | atr-miR167 | MIMAT0033919 | |
| Asparagus officinalis | aof-miR167c | MIMAT0049779 | |
| Citrus sinensis | csi-miR167a-5p | MIMAT0018498 | |
| Citrus sinensis | csi-miR167c-5p | MIMAT0018453 | |
| Cucumis melo | cme-miR167d | MIMAT0026163 | |
| Cucumis melo | cme-miR167f | MIMAT0026164 | |
| View More Rows | |||
| Digitalis purpurea | dpr-miR167c | MIMAT0023533 | |
| Glycine max | gma-miR167c | MIMAT0007355 | |
| Glycine max | gma-miR167j | MIMAT0021677 | |
| Glycine soja | gso-miR167a | MIMAT0016351 | |
| Linum usitatissimum | lus-miR167f | MIMAT0027175 | |
| Linum usitatissimum | lus-miR167g | MIMAT0027176 | |
| Linum usitatissimum | lus-miR167i | MIMAT0027190 | |
| Lotus japonicus | lja-miR167 | MIMAT0010087 | |
| Manihot esculenta | mes-miR167a | MIMAT0024413 | |
| Medicago truncatula | mtr-miR167b-5p | MIMAT0022233 | |
| Populus trichocarpa | ptc-miR167e | MIMAT0001945 | |
| Rice | osa-miR167d-5p | MIMAT0001039 | |
| Rice | osa-miR167e-5p | MIMAT0001040 | |
| Rice | osa-miR167f | MIMAT0001041 | |
| Rice | osa-miR167g | MIMAT0001042 | |
| Rice | osa-miR167h-5p | MIMAT0001043 | |
| Rice | osa-miR167i-5p | MIMAT0001044 | |
| Rice | osa-miR167j | MIMAT0001086 | |
| Saccharum officinarum | sof-miR167a | MIMAT0001663 | |
| Saccharum officinarum | sof-miR167b | MIMAT0001664 | |
| Saccharum sp. | ssp-miR167b | MIMAT0020293 | |
| Sorghum bicolor | sbi-miR167c | MIMAT0001451 | |
| Sorghum bicolor | sbi-miR167d | MIMAT0001447 | |
| Sorghum bicolor | sbi-miR167e | MIMAT0001450 | |
| Sorghum bicolor | sbi-miR167f | MIMAT0001448 | |
| Sorghum bicolor | sbi-miR167g | MIMAT0001449 | |
| Sorghum bicolor | sbi-miR167h | MIMAT0011331 | |
| Vitis vinifera | vvi-miR167a | MIMAT0005670 | |
| Zea mays | zma-miR167e-5p | MIMAT0001721 | |
| Zea mays | zma-miR167f-5p | MIMAT0001722 | |
| Zea mays | zma-miR167g-5p | MIMAT0001723 | |
| Zea mays | zma-miR167h-5p | MIMAT0001724 | |
| Zea mays | zma-miR167i-5p | MIMAT0001725 | |
| Zea mays | zma-miR167j-5p | MIMAT0013987 | |