Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 003188_mat
          Assay Name: bfl-miR-124
          miRBase Accession Number: MI0000443
          miRBase Version: v22.1
          Chromosome Location: Chr.8: 9903388 - 9903472 [-] on Build GRCh38
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCCAA
          Species: Human, Branchiostoma belcheri, Branchiostoma floridae, Brugia malayi, Ciona intestinalis, Ciona savignyi, Columba livia, Cricetulus griseus, Dasypus novemcinctus, Fugu rubripes, Guinea pig, Haemonchus contortus, Heliconius melpomene, Paralichthys olivaceus, Patiria miniata, Pteropus alecto, Python bivittatus, Saccoglossus kowalevskii, Tetraodon nigroviridis, Triops cancriformis, Xenoturbella bocki, Zebrafish
          Product Type: TaqMan™ MicroRNA Assay
          Assay ID 003188_mat
          Size
          Availability Made To Order
          Catalog # 4440886
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000021, mir-124

          Mature miRNA Sequence:

          UAAGGCACGCGGUGAAUGCCAA

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-124-3p MIMAT0000422
          Branchiostoma belcheri bbe-miR-124-3p MIMAT0031578
          Branchiostoma floridae bfl-miR-124-3p MIMAT0009482
          Brugia malayi bma-miR-124 MIMAT0014116
          Ciona intestinalis cin-miR-124-3p MIMAT0006100
          Ciona savignyi csa-miR-124 MIMAT0006126
          View More Rows
          Columba livia cli-miR-124-3p MIMAT0038494
          Cricetulus griseus cgr-miR-124 MIMAT0023740
          Dasypus novemcinctus dno-miR-124-3p MIMAT0047613
          Fugu rubripes fru-miR-124 MIMAT0002896
          Guinea pig cpo-miR-124-3p MIMAT0046964
          Haemonchus contortus hco-miR-124 MIMAT0023338
          Heliconius melpomene hme-miR-124 MIMAT0024937
          Paralichthys olivaceus pol-miR-124-3p MIMAT0025431
          Patiria miniata pmi-miR-124-3p MIMAT0032157
          Pteropus alecto pal-miR-124-3p MIMAT0039837
          Python bivittatus pbv-miR-124a-3p MIMAT0038914
          Saccoglossus kowalevskii sko-miR-124-3p MIMAT0009625
          Tetraodon nigroviridis tni-miR-124 MIMAT0002897
          Triops cancriformis tcf-miR-124-3p MIMAT0041369
          Xenoturbella bocki xbo-miR-124 MIMAT0020120
          Zebrafish dre-miR-124-3p MIMAT0001819

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-124-1
          Stem-loop Accession # MI0000443
          Stem-loop Sequence
          AGGCCUCUCUCUCCGUGUUCACAGCGGACCUUGAUUUAAAUGUCCAUACAAUUAAGGCACGCGGUGAAUGCCAAGAAUGGGGCUG
          Chromosome Location Chr. 8 - 9903388 - 9903472 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000422
          miRBase ID: hsa-miR-124-3p
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCCAA
          Chromosome Location: Chr. 8 - 9903388 - 9903472 [-] on Build GRCh38

          Stem-loop ID hsa-mir-124-2
          Stem-loop Accession # MI0000444
          Stem-loop Sequence
          AUCAAGAUUAGAGGCUCUGCUCUCCGUGUUCACAGCGGACCUUGAUUUAAUGUCAUACAAUUAAGGCACGCGGUGAAUGCCAAGAGCGGAGCCUACGGCUGCACUUGAA
          Chromosome Location Chr. 8 - 64379149 - 64379257 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000422
          miRBase ID: hsa-miR-124-3p
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCCAA
          Chromosome Location: Chr. 8 - 9903388 - 9903472 [-] on Build GRCh38

          Stem-loop ID hsa-mir-124-3
          Stem-loop Accession # MI0000445
          Stem-loop Sequence
          UGAGGGCCCCUCUGCGUGUUCACAGCGGACCUUGAUUUAAUGUCUAUACAAUUAAGGCACGCGGUGAAUGCCAAGAGAGGCGCCUCC
          Chromosome Location Chr. 20 - 63178500 - 63178586 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000422
          miRBase ID: hsa-miR-124-3p
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCCAA
          Chromosome Location: Chr. 8 - 9903388 - 9903472 [-] on Build GRCh38

          Branchiostoma belcheri

          Stem-loop ID bbe-mir-124
          Stem-loop Accession # MI0026176
          Stem-loop Sequence
          CGGCAAAUCUCGGCUUGGCUCUUAGUGUUCACGGCGGUCCUUAAUUUAACAUUGUGCAAUUAAGGCACGCGGUGAAUGCCAAGAACAGGGCCAAAUCCUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0031578
          miRBase ID: bbe-miR-124-3p
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCCAA
          Chromosome Location: NA

          Branchiostoma floridae

          Stem-loop ID bfl-mir-124
          Stem-loop Accession # MI0010026
          Stem-loop Sequence
          CGGCAAAUCUCGGCUUGGCUCUUAGUGUUCACGGCGGUCCUUAAUUUAACAUUGUGCAAUUAAGGCACGCGGUGAAUGCCAAGAACAGGGCCAAAUCCUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009482
          miRBase ID: bfl-miR-124-3p
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCCAA
          Chromosome Location: NA

          Brugia malayi

          Stem-loop ID bma-mir-124
          Stem-loop Accession # MI0013342
          Stem-loop Sequence
          CAAAGGACGUCUCGCUUUCACCGGUGACCUUGGUUUGUUAUGUUUAGUACUAAGGCACGCGGUGAAUGCCAAACGUUCUCUUGCUAUACCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014116
          miRBase ID: bma-miR-124
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCCAA
          Chromosome Location: NA

          Ciona intestinalis

          Stem-loop ID cin-mir-124-1
          Stem-loop Accession # MI0007164
          Stem-loop Sequence
          UAUGUUCGGUAGUAUUUAUUGUGGACCUUGCUGUGUGACGUCACAAUAAGGCACGCGGUGAAUGCCAACGAAUUUUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006100
          miRBase ID: cin-miR-124-3p
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCCAA
          Chromosome Location: NA

          Stem-loop ID cin-mir-124-2
          Stem-loop Accession # MI0007165
          Stem-loop Sequence
          CGUAAUAUUUAAAGGGCGCUUAUGAGUCGUGUUUACUGUGGACCUUGCUGUGUGACGUCACAAUAAGGCACGCGGUGAAUGCCAAUCAAAAGUCGCCGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006100
          miRBase ID: cin-miR-124-3p
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCCAA
          Chromosome Location: NA

          Ciona savignyi

          Stem-loop ID csa-mir-124-1
          Stem-loop Accession # MI0007192
          Stem-loop Sequence
          UUAUAAUCGACGUCAUAGCCAAGUUCGAUCGUGUUUAUUGUGGACCUUGCCGUGUGACGUCACAGUAAGGCACGCGGUGAAUGCCAACGAACUCGCGCUGCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006126
          miRBase ID: csa-miR-124
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCCAA
          Chromosome Location: NA

          Stem-loop ID csa-mir-124-2
          Stem-loop Accession # MI0007193
          Stem-loop Sequence
          GUGACGUAAAAUCAAUAAUGCGCUUCAUCCGCGUUUGCUGCGGACCUUCUUGUGUGACAUCACAAUAAGGCACGCGGUGAAUGCCAAUGAAACGUCAUCACU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006126
          miRBase ID: csa-miR-124
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCCAA
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-mir-124-1
          Stem-loop Accession # MI0029954
          Stem-loop Sequence
          CCAAGGCUCUGACUCUCCGUGUUCACAGCGGACCUUGAUUUAAUGUCAUACAAUUAAGGCACGCGGUGAAUGCCAAGAGCGGAUCCUCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038494
          miRBase ID: cli-miR-124-3p
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCCAA
          Chromosome Location: NA

          Stem-loop ID cli-mir-124-2
          Stem-loop Accession # MI0029955
          Stem-loop Sequence
          CCCGAGCCCCUCUCUGCGUGUUCACAGCGGACCUUGAUUUAAUGUCCAUACAAUUAAGGCACGCGGUGAAUGCCAAGAGAGGAGCCUCACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038494
          miRBase ID: cli-miR-124-3p
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCCAA
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-124
          Stem-loop Accession # MI0020384
          Stem-loop Sequence
          ACUCUGCCCUCCGUGUUCACAGCGGACCUUGAUUUAAUGUCAUACAAUUAAGGCACGCGGUGAAUGCCAAGAGCGGAGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023740
          miRBase ID: cgr-miR-124
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCCAA
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-124-1
          Stem-loop Accession # MI0038964
          Stem-loop Sequence
          CGUGUUCACAGCGGACCUUGAUUUAAUGUCAUACAAUUAAGGCACGCGGUGAAUGCCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047613
          miRBase ID: dno-miR-124-3p
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCCAA
          Chromosome Location: NA

          Stem-loop ID dno-mir-124-2
          Stem-loop Accession # MI0038965
          Stem-loop Sequence
          CGUGUUCACAGCGGACCUUGAUUUAAUGUCUACACAAUUAAGGCACGCGGUGAAUGCCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047613
          miRBase ID: dno-miR-124-3p
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCCAA
          Chromosome Location: NA

          Fugu rubripes

          Stem-loop ID fru-mir-124-1
          Stem-loop Accession # MI0003287
          Stem-loop Sequence
          GGUUGUGUCUCUCCGUGUUCACAGCGGACCUUGAUUUAAUGUCUUACAAUUAAGGCACGCGGUGAAUGCCAAGAGAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002896
          miRBase ID: fru-miR-124
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCCAA
          Chromosome Location: NA

          Stem-loop ID fru-mir-124-2
          Stem-loop Accession # MI0003354
          Stem-loop Sequence
          CUGGUCUCUCCUCGUGUUCACAGCGGACCUUGAUUUAAAUGUCCAUACAAUUAAGGCACGCGGUGAAUGCCAAGAGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002896
          miRBase ID: fru-miR-124
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCCAA
          Chromosome Location: NA

          Stem-loop ID fru-mir-124-3
          Stem-loop Accession # MI0003211
          Stem-loop Sequence
          GGUUUGAGCUCUUUGUGUUCACAGUGGACCUUGAUUUAAUUUCAAUACAAUUAAGGCACGCGGUGAAUGCCAAGAGAGAAGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002896
          miRBase ID: fru-miR-124
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCCAA
          Chromosome Location: NA

          Stem-loop ID fru-mir-124-4
          Stem-loop Accession # MI0031746
          Stem-loop Sequence
          GGUUGUGUCUCUCCGUGUUCACAGCGGACCUUGAUUUAAUGUCUUACAAUUAAGGCACGCGGUGAAUGCCAAGAGAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002896
          miRBase ID: fru-miR-124
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCCAA
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-124-1
          Stem-loop Accession # MI0038622
          Stem-loop Sequence
          CGUGUUCACAGCGGACCUUGAUUUAAAUGUCCAUACAAUUAAGGCACGCGGUGAAUGCCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046964
          miRBase ID: cpo-miR-124-3p
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCCAA
          Chromosome Location: NA

          Stem-loop ID cpo-mir-124-2
          Stem-loop Accession # MI0038623
          Stem-loop Sequence
          CGUGUUCACAGCGGACCUUGAUUUAAUGUCAUACAAUUAAGGCACGCGGUGAAUGCCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046964
          miRBase ID: cpo-miR-124-3p
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCCAA
          Chromosome Location: NA

          Haemonchus contortus

          Stem-loop ID hco-mir-124
          Stem-loop Accession # MI0020014
          Stem-loop Sequence
          CCGUAUCGACGCUGGAUCGCUUCGCUUUCAUCGGUGACUUUAGUGUGAAAUGCAACUAAGGCACGCGGUGAAUGCCAAGCGAAUAAAGCGUUUCUUCCUCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023338
          miRBase ID: hco-miR-124
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCCAA
          Chromosome Location: NA

          Heliconius melpomene

          Stem-loop ID hme-mir-124
          Stem-loop Accession # MI0021732
          Stem-loop Sequence
          GAGCAUUAAGAACACAGUCCACCUCCUCGCGUUCACUGCGCAGCCAUAUGUGUUCCUAUCCAUCAUAAGGCACGCGGUGAAUGCCAAGAGCGGGCUCAAGCGCCGAUUGCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024937
          miRBase ID: hme-miR-124
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCCAA
          Chromosome Location: NA

          Paralichthys olivaceus

          Stem-loop ID pol-mir-124
          Stem-loop Accession # MI0022198
          Stem-loop Sequence
          CGUGUUCACAGCGGACCUUGAUUUAAUGUCCAUACAAUUAAGGCACGCGGUGAAUGCCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0025431
          miRBase ID: pol-miR-124-3p
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCCAA
          Chromosome Location: NA

          Patiria miniata

          Stem-loop ID pmi-mir-124
          Stem-loop Accession # MI0025105
          Stem-loop Sequence
          AAUAUUUUGUAUCCAGUUCUUAGUGUUCACUGCGAUCCUUGAUUUACACACAACGUGGGCAAUUAAGGCACGCGGUGAAUGCCAAGAGUCUGGAAUCCAUACACAUCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032157
          miRBase ID: pmi-miR-124-3p
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCCAA
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-124
          Stem-loop Accession # MI0032354
          Stem-loop Sequence
          CGUGUUCACAGCGGACCUUGAUUUAAUGUCAUACAAUUAAGGCACGCGGUGAAUGCCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039837
          miRBase ID: pal-miR-124-3p
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCCAA
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-mir-124a-1
          Stem-loop Accession # MI0030212
          Stem-loop Sequence
          CGAGGCUCUGUCUCUCCGUGUUCACAGCGGACCUUGAUUUAAUGUCAUACAAUUAAGGCACGCGGUGAAUGCCAAGAGCAGGGCUUGAAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038914
          miRBase ID: pbv-miR-124a-3p
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCCAA
          Chromosome Location: NA

          Stem-loop ID pbv-mir-124a-2
          Stem-loop Accession # MI0030213
          Stem-loop Sequence
          CCUGGGCCCUUCUCUCCGUGUUCACAGCGGACCUUGAUUUAAUGUCCAUACAAUUAAGGCACGCGGUGAAUGCCAAGAGACCGGUUCCCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038914
          miRBase ID: pbv-miR-124a-3p
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCCAA
          Chromosome Location: NA

          Stem-loop ID pbv-mir-124a-3
          Stem-loop Accession # MI0030214
          Stem-loop Sequence
          CUUGGCUUAAGCCCUUCGUGUUCACAGCGGACCUUGAUUUAAUUUCGAUACAAUUAAGGCACGCGGUGAAUGCCAAGAGAGAAGCCACGUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038914
          miRBase ID: pbv-miR-124a-3p
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCCAA
          Chromosome Location: NA

          Saccoglossus kowalevskii

          Stem-loop ID sko-mir-124
          Stem-loop Accession # MI0010162
          Stem-loop Sequence
          GGAUGGUUAUGUCCUGUUCUAAGUGUUCACAGCGGUCCUUGAUGUUCCACACAACGCAAUUAAGGCACGCGGUGAAUGCCAAGAACAAAGAUAUAUAAAAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009625
          miRBase ID: sko-miR-124-3p
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCCAA
          Chromosome Location: NA

          Tetraodon nigroviridis

          Stem-loop ID tni-mir-124-1
          Stem-loop Accession # MI0003288
          Stem-loop Sequence
          CUCUCCGUGUUCACAGCGGACCUUGAUUUAAUGUCUUACAAUUAAGGCACGCGGUGAAUGCCAAGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002897
          miRBase ID: tni-miR-124
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCCAA
          Chromosome Location: NA

          Stem-loop ID tni-mir-124-2
          Stem-loop Accession # MI0003355
          Stem-loop Sequence
          GCCUCUCCUCGUGUUCACAGCGGACCUUGAUUUAAAUGUCCAUACAAUUAAGGCACGCGGUGAAUGCCAAGAGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002897
          miRBase ID: tni-miR-124
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCCAA
          Chromosome Location: NA

          Stem-loop ID tni-mir-124-3
          Stem-loop Accession # MI0003212
          Stem-loop Sequence
          GGUUUGAGCUCUUUGUGUUCACAGUGGACCUUGAUUUAAUUUCAAUACAAUUAAGGCACGCGGUGAAUGCCAAGAGAGAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002897
          miRBase ID: tni-miR-124
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCCAA
          Chromosome Location: NA

          Triops cancriformis

          Stem-loop ID tcf-mir-124
          Stem-loop Accession # MI0033209
          Stem-loop Sequence
          CGUGUUCACUGUUGGCCUUGAUGUGGAUAUUGUCAUAAGGCACGCGGUGAAUGCCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0041369
          miRBase ID: tcf-miR-124-3p
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCCAA
          Chromosome Location: NA

          Xenoturbella bocki

          Stem-loop ID xbo-mir-124
          Stem-loop Accession # MI0017686
          Stem-loop Sequence
          GAAGGAGAGAGUUUGGUAGUGCUUCGUGUUCACAGCGAGGUCCUUGAUUCGUGUUACAAUAAUUAAGGCACGCGGUGAAUGCCAAGAACUACCGGGCCGAAAUCAUCGCUGGAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0020120
          miRBase ID: xbo-miR-124
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCCAA
          Chromosome Location: NA

          Zebrafish

          Stem-loop ID dre-mir-124-1
          Stem-loop Accession # MI0001966
          Stem-loop Sequence
          GGCUCUCGCUGUACGUGUUCACAGUGGACCUUGAUUUAUUGUAUUUCAAUUAAGGCACGCGGUGAAUGCCAACAGCACAGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001819
          miRBase ID: dre-miR-124-3p
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCCAA
          Chromosome Location: NA

          Stem-loop ID dre-mir-124-2
          Stem-loop Accession # MI0001967
          Stem-loop Sequence
          CCUGCUUUUCUUCGUGUUCACAGCGGACCUUGAUUUAAAUGUCCAUACAAUUAAGGCACGCGGUGAAUGCCAAGAGAGAUGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001819
          miRBase ID: dre-miR-124-3p
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCCAA
          Chromosome Location: NA

          Stem-loop ID dre-mir-124-3
          Stem-loop Accession # MI0001968
          Stem-loop Sequence
          GGCUCUGUGGGAUUUCAGACUCUGGCUUUCCGUGUUCACAGCGGACCUUGAUUUAAUGUCUUACAAUUAAGGCACGCGGUGAAUGCCAAGAGCGGAGCCUUUUAACAUCAGCAGGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001819
          miRBase ID: dre-miR-124-3p
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCCAA
          Chromosome Location: NA

          Stem-loop ID dre-mir-124-4
          Stem-loop Accession # MI0001969
          Stem-loop Sequence
          GGUUUUUGCUCUUUGUGUUCACAGUGGACCUUGAUUUAAUUUCAAUACAAUUAAGGCACGCGGUGAAUGCCAAGAGAGAAGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001819
          miRBase ID: dre-miR-124-3p
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCCAA
          Chromosome Location: NA

          Stem-loop ID dre-mir-124-5
          Stem-loop Accession # MI0001970
          Stem-loop Sequence
          GGGUUUUGCUCGUGCGUUCUUUUUGAGUUCUCGCUCUGCGUGUUCACAGCGGACCUUGAUUUAAUGUCCAUACAAUUAAGGCACGCGGUGAAUGCCAAGAGAAGAAUCUCUCCAGCAACGAGUUUGCGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001819
          miRBase ID: dre-miR-124-3p
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCCAA
          Chromosome Location: NA

          Stem-loop ID dre-mir-124-6
          Stem-loop Accession # MI0001971
          Stem-loop Sequence
          GGGUGGUGACACAGGCCCGCCACUCUGCGUGUUCACGGCGGACCUUGAUUUAAUAUCCAUACAAUUAAGGCACGCGGUGAAUGCCAAGAGAGGGGUCUUAAAACGACAAACCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001819
          miRBase ID: dre-miR-124-3p
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCCAA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303081_pri
          TaqMan™ Pri-miRNA Assay : Hs03303077_pri
          TaqMan™ Pri-miRNA Assay : Hs03303090_pri
          mirVana® miRNA inhibitor : MH10060
          mirVana® miRNA mimic : MC10060
          TaqMan™ Advanced miRNA Assay : 480901_mir


          miRBase Alias:

          Branchiostoma floridae: bfl-miR-124 (v18)
          Chryseobacterium indologenes: cin-miR-124 (v19)
          Danio rerio: dre-miR-124 (v20)
          Streptococcus koreensis: sko-miR-124 (v18)

          Back To Top

          Related Products

          • TaqMan® Universal Master Mix II, with UNG
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® MicroRNA Reverse Transcription Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline