Search
Search

| Assay Name: | oan-let-7d* |
|---|---|
| miRBase Accession Number: | MI0029888 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CUAUACAACCUGCUGCCUUUCU |
| Species: | Chrysemys picta, Columba livia, Ophiophagus hannah, Ornithorhynchus anatinus, Python bivittatus, Taeniopygia guttata |
| Product Type: | TaqMan™ MicroRNA Assay |
Mature miRNA Sequence: | CUAUACAACCUGCUGCCUUUCU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Chrysemys picta | cpi-let-7d-3p | MIMAT0037603 |
| Columba livia | cli-let-7d-3p | MIMAT0038382 |
| Ophiophagus hannah | oha-let-7d-3p | MIMAT0036637 |
| Ornithorhynchus anatinus | oan-let-7d-3p | MIMAT0007235 |
| Python bivittatus | pbv-let-7d-3p | MIMAT0038802 |
| Taeniopygia guttata | tgu-let-7d-3p | MIMAT0026992 |