Search
Search

| Assay Name: | lmi-miR-210 |
|---|---|
| miRBase Accession Number: | MI0000376 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CUUGUGCGUGUGACAGCGGCUAU |
| Species: | Drosophila melanogaster, Locusta migratoria, Manduca sexta |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000086, mir-210 |
Mature miRNA Sequence: | CUUGUGCGUGUGACAGCGGCUAU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Drosophila melanogaster | dme-miR-210-3p | MIMAT0000355 |
| Locusta migratoria | lmi-miR-210-3p | MIMAT0010143 |
| Manduca sexta | mse-miR-210 | MIMAT0024420 |