Search
Search

| Assay Name: | bfl-miR-29a |
|---|---|
| miRBase Accession Number: | MI0026231 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UAGCACCAUAUGAAAUCAGUUAU |
| Species: | Branchiostoma belcheri, Branchiostoma floridae |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000009, mir-29 |
Mature miRNA Sequence: | UAGCACCAUAUGAAAUCAGUUAU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Branchiostoma belcheri | bbe-miR-29a-3p | MIMAT0031663 |
| Branchiostoma floridae | bfl-miR-29a-3p | MIMAT0009471 |