Search
Search

| Assay Name: | ppt-miR1048-5p |
|---|---|
| miRBase Accession Number: | MI0006007 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UAGAACAUGAGUGUAGACGAC |
| Species: | Physcomitrella patens |
| Product Type: | TaqMan™ MicroRNA Assay |
Mature miRNA Sequence: | UAGAACAUGAGUGUAGACGAC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Physcomitrella patens | ppt-miR1048-5p | MIMAT0005163 |