Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 006765_mat
          Assay Name: oan-miR-150
          miRBase Accession Number: MI0006840
          miRBase Version: v22.1
          Chromosome Location: -
          Mature miRNA Sequence: UCUCCCAACCCUUGUACCAGAG
          Species: Ornithorhynchus anatinus, Xenopus tropicalis
          Product Type: TaqMan™ MicroRNA Assay
          Assay ID 006765_mat
          Size
          Availability Made To Order
          Catalog # 4440886
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000197, mir-150

          Mature miRNA Sequence:

          UCUCCCAACCCUUGUACCAGAG

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Ornithorhynchus anatinus oan-miR-150-5p MIMAT0007090
          Xenopus tropicalis xtr-miR-150 MIMAT0003606

          Stem-loop Details



          Ornithorhynchus anatinus

          Stem-loop ID oan-mir-150
          Stem-loop Accession # MI0006840
          Stem-loop Sequence
          CGCCCCUCGGCGCCCCCUCUCCCAACCCUUGUACCAGAGCAUAAACUGAACCCCUGGUACGGGGCCGUGGGAGAGGGGGAGCCGGGGCCCCGCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0007090
          miRBase ID: oan-miR-150-5p
          Mature miRNA Sequence: UCUCCCAACCCUUGUACCAGAG
          Chromosome Location: NA

          Xenopus tropicalis

          Stem-loop ID xtr-mir-150
          Stem-loop Accession # MI0004846
          Stem-loop Sequence
          CUCUCUCCCAACCCUUGUACCAGAGUGAUAAUGGGAACUCUGGUACAGAGGAUGGCUGAAAGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003606
          miRBase ID: xtr-miR-150
          Mature miRNA Sequence: UCUCCCAACCCUUGUACCAGAG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          mirVana® miRNA inhibitor : MH11630
          mirVana® miRNA mimic : MC11630


          miRBase Alias:

          Ochrobactrum anthropi: oan-miR-150 (v18)

          Back To Top

          Related Products

          • TaqMan® Universal Master Mix II, with UNG
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® MicroRNA Reverse Transcription Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline