Search
Search

| Assay Name: | oan-miR-383* |
|---|---|
| miRBase Accession Number: | MI0038763 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CCACAACACUGCCUGGUCAGA |
| Species: | Guinea pig, Ornithorhynchus anatinus |
| Product Type: | TaqMan™ MicroRNA Assay |
Mature miRNA Sequence: | CCACAACACUGCCUGGUCAGA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Guinea pig | cpo-miR-383-3p | MIMAT0047230 |
| Ornithorhynchus anatinus | oan-miR-383-3p | MIMAT0007190 |