Search
Search

| Assay Name: | aga-miR-927 |
|---|---|
| miRBase Accession Number: | MI0005843 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UUUAGAAUUCCUACGCUUUACC |
| Species: | Drosophila melanogaster, Acyrthosiphon pisum, Anopheles gambiae, Bombyx mori, Dinoponera quadriceps, Drosophila pseudoobscura, Drosophila simulans, Drosophila virilis, Heliconius melpomene, Polistes canadensis |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000452, mir-927 |
Mature miRNA Sequence: | UUUAGAAUUCCUACGCUUUACC |
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Drosophila melanogaster | dme-miR-927-5p | MIMAT0005501 | |
| Acyrthosiphon pisum | api-miR-927 | MIMAT0014740 | |
| Anopheles gambiae | aga-miR-927 | MIMAT0010102 | |
| Bombyx mori | bmo-miR-927-5p | MIMAT0009157 | |
| Dinoponera quadriceps | dqu-miR-927a-5p | MIMAT0045745 | |
| Drosophila pseudoobscura | dps-miR-927-5p | MIMAT0012269 | |
| View More Rows | |||
| Drosophila simulans | dsi-miR-927-5p | MIMAT0012483 | |
| Drosophila virilis | dvi-miR-927-5p | MIMAT0035570 | |
| Heliconius melpomene | hme-miR-927 | MIMAT0024990 | |
| Polistes canadensis | pca-miR-927a-5p | MIMAT0045911 | |