Search
Search

| Assay Name: | ppy-miR-615-5p |
|---|---|
| miRBase Accession Number: | MI0015100 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GGGGGUUCCCCGGUGCUCGGAUC |
| Species: | Pongo pygmaeus |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000342, mir-615 |
Mature miRNA Sequence: | GGGGGUUCCCCGGUGCUCGGAUC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Pongo pygmaeus | ppy-miR-615-5p | MIMAT0016056 |