Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 241456_mat
          Assay Name: hsa-miR-3198
          miRBase Accession Number: MI0014246
          miRBase Version: v22.1
          Chromosome Location: Chr.22: 17764180 - 17764259 [-] on Build GRCh38
          Mature miRNA Sequence: GUGGAGUCCUGGGGAAUGGAGA
          Species: Human
          Product Type: TaqMan™ MicroRNA Assay
          Assay ID 241456_mat
          Size
          Availability Inventoried
          Catalog # 4427975
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0001216, mir-3198

          Mature miRNA Sequence:

          GUGGAGUCCUGGGGAAUGGAGA

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-3198 MIMAT0015083

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-3198-1
          Stem-loop Accession # MI0014246
          Stem-loop Sequence
          GACUGUGCUCUCACUGUUCACCCAGCACUAGCAGUACCAGACGGUUCUGUGGAGUCCUGGGGAAUGGAGAGAGCACAGUC
          Chromosome Location Chr. 22 - 17764180 - 17764259 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0015083
          miRBase ID: hsa-miR-3198
          Mature miRNA Sequence: GUGGAGUCCUGGGGAAUGGAGA
          Chromosome Location: Chr. 22 - 17764180 - 17764259 [-] on Build GRCh38

          Stem-loop ID hsa-mir-3198-2
          Stem-loop Accession # MI0017335
          Stem-loop Sequence
          GACUCUGCUCUCACUGUUCACCCAGCACUAGCAGUACCAGAUGGUUCUGUGGAGUCCUGGGGAAUGGAGAGAGCACAGUC
          Chromosome Location Chr. 12 - 54231397 - 54231476 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0015083
          miRBase ID: hsa-miR-3198
          Mature miRNA Sequence: GUGGAGUCCUGGGGAAUGGAGA
          Chromosome Location: Chr. 22 - 17764180 - 17764259 [-] on Build GRCh38


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs04227606_pri
          TaqMan™ Pri-miRNA Assay : Hs04257655_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM16633
          Ambion® Pre-miR™ miRNA Precursor : PM16633
          mirVana® miRNA inhibitor : MH16633
          mirVana® miRNA mimic : MC16633
          TaqMan™ Advanced miRNA Assay : 478823_mir

          Back To Top

          Related Products

          • TaqMan® Universal Master Mix II, with UNG
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® MicroRNA Reverse Transcription Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline