Search
Search

| Assay Name: | hsa-miR-3154 |
|---|---|
| miRBase Accession Number: | MI0014182 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CAGAAGGGGAGUUGGGAGCAGA |
| Species: | Human, Cricetulus griseus |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0001387, mir-3154 |
Mature miRNA Sequence: | CAGAAGGGGAGUUGGGAGCAGA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-3154 | MIMAT0015028 |
| Cricetulus griseus | cgr-miR-3154 | MIMAT0041139 |