Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 242639_mat
          Assay Name: bta-miR-2904
          miRBase Accession Number: MI0013080
          miRBase Version: v22.1
          Chromosome Location: -
          Mature miRNA Sequence: GGGAGCCUCGGUUGGCCUC
          Species: Bovine
          Product Type: TaqMan™ MicroRNA Assay
          Assay ID 242639_mat
          Size
          Availability Made To Order
          Catalog # 4440886
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000872, mir-2904

          Mature miRNA Sequence:

          GGGAGCCUCGGUUGGCCUC

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Bovine bta-miR-2904 MIMAT0013862

          Stem-loop Details



          Bovine

          Stem-loop ID bta-mir-2904-1
          Stem-loop Accession # MI0013080
          Stem-loop Sequence
          ACGGCAGCGCCGCGGGAGCCUCGGUUGGCCUCGGAUAGCCGGUCCCCCGCCGUCCCCGCCGGCGGGCCGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013862
          miRBase ID: bta-miR-2904
          Mature miRNA Sequence: GGGAGCCUCGGUUGGCCUC
          Chromosome Location: NA

          Stem-loop ID bta-mir-2904-2
          Stem-loop Accession # MI0013081
          Stem-loop Sequence
          ACGGCAGCGCCGCGGGAGCCUCGGUUGGCCUCGGAUAGCCGGUCCCCCGCCGUCCCCGCCGGCGGGCCGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013862
          miRBase ID: bta-miR-2904
          Mature miRNA Sequence: GGGAGCCUCGGUUGGCCUC
          Chromosome Location: NA

          Stem-loop ID bta-mir-2904-3
          Stem-loop Accession # MI0013082
          Stem-loop Sequence
          ACGGCAGCGCCGCGGGAGCCUCGGUUGGCCUCGGAUAGCCGGUCCCCCGCCGUCCCCGCCGGCGGGCCGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013862
          miRBase ID: bta-miR-2904
          Mature miRNA Sequence: GGGAGCCUCGGUUGGCCUC
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          mirVana® miRNA inhibitor : MH17288
          mirVana® miRNA mimic : MC17288

          Back To Top

          Related Products

          • TaqMan® Universal Master Mix II, with UNG
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® MicroRNA Reverse Transcription Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline