Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 245467_mat
          Assay Name: hsa-miR-1226
          miRBase Accession Number: MI0006313
          miRBase Version: v22.1
          Chromosome Location: Chr.3: 47849555 - 47849629 [+] on Build GRCh38
          Mature miRNA Sequence: UCACCAGCCCUGUGUUCCCUAG
          Species: Human, Pan troglodytes, Pongo pygmaeus, Rhesus monkey
          Product Type: TaqMan™ MicroRNA Assay
          Assay ID 245467_mat
          Size
          Availability Inventoried
          Catalog # 4427975
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000444, mir-1226

          Mature miRNA Sequence:

          UCACCAGCCCUGUGUUCCCUAG

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-1226-3p MIMAT0005577
          Pan troglodytes ptr-miR-1226 MIMAT0005578
          Pongo pygmaeus ppy-miR-1226 MIMAT0016162
          Rhesus monkey mml-miR-1226 MIMAT0005579

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-1226
          Stem-loop Accession # MI0006313
          Stem-loop Sequence
          GUGAGGGCAUGCAGGCCUGGAUGGGGCAGCUGGGAUGGUCCAAAAGGGUGGCCUCACCAGCCCUGUGUUCCCUAG
          Chromosome Location Chr. 3 - 47849555 - 47849629 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0005577
          miRBase ID: hsa-miR-1226-3p
          Mature miRNA Sequence: UCACCAGCCCUGUGUUCCCUAG
          Chromosome Location: Chr. 3 - 47849555 - 47849629 [+] on Build GRCh38

          Pan troglodytes

          Stem-loop ID ptr-mir-1226
          Stem-loop Accession # MI0006314
          Stem-loop Sequence
          GUGAGGGCAUGCAGGCCUGGAUGGGGCAGCUGGGAUGGUCCAAAAGGGUGGCCUCACCAGCCCUGUGUUCCCUAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0005578
          miRBase ID: ptr-miR-1226
          Mature miRNA Sequence: UCACCAGCCCUGUGUUCCCUAG
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-1226
          Stem-loop Accession # MI0015195
          Stem-loop Sequence
          GUGAGGGCAUGCAGGCCUGGAUGGGGCAGCUGGGAUGGUCCAAAAGGGUGGCCUCACCAGCCCUGUGUUCCCUAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0016162
          miRBase ID: ppy-miR-1226
          Mature miRNA Sequence: UCACCAGCCCUGUGUUCCCUAG
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-1226
          Stem-loop Accession # MI0006315
          Stem-loop Sequence
          GUGAGGGCAUGCAGGCCUGGAUGGGGCAGCUGGGAUGGUCCAAAAGGGUGGCCUCACCAGCCCUGUGUUCCCUAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0005579
          miRBase ID: mml-miR-1226
          Mature miRNA Sequence: UCACCAGCCCUGUGUUCCCUAG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03305379_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM13533
          Ambion® Pre-miR™ miRNA Precursor : PM13533
          mirVana® miRNA inhibitor : MH13533
          mirVana® miRNA mimic : MC13533
          TaqMan™ Advanced miRNA Assay : 478640_mir


          miRBase Alias:

          Human: hsa-miR-1226 (v17)

          Back To Top

          Related Products

          • TaqMan® Universal Master Mix II, with UNG
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® MicroRNA Reverse Transcription Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline