Search
Search

| Assay Name: | mmu-miR-429* |
|---|---|
| miRBase Accession Number: | MI0001642 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GUCUUACCAGACAUGGUUAGA |
| Species: | Mouse |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000019, mir-8 |
Mature miRNA Sequence: | GUCUUACCAGACAUGGUUAGA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-429-5p | MIMAT0017178 |