Search
Search

| Assay Name: | ssc-miR-708-3p |
|---|---|
| miRBase Accession Number: | MI0038838 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CAACUAGACUGUGAGCUUCUAGA |
| Species: | Guinea pig, Pig, Pteropus alecto, Rabbit, Rhesus monkey |
| Product Type: | TaqMan™ MicroRNA Assay |
Mature miRNA Sequence: | CAACUAGACUGUGAGCUUCUAGA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Guinea pig | cpo-miR-708-3p | MIMAT0047378 |
| Pig | ssc-miR-708-3p | MIMAT0017383 |
| Pteropus alecto | pal-miR-708-3p | MIMAT0040004 |
| Rabbit | ocu-miR-708-3p | MIMAT0048562 |
| Rhesus monkey | mml-miR-708-3p | MIMAT0026952 |