Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 462575_mat
          Assay Name: hsa-miR-1246
          miRBase Accession Number: MI0006381
          miRBase Version: v20
          Chromosome Location: Chr.2: 177465708 - 177465780 [-] on Build 37
          Mature miRNA Sequence: AAUGGAUUUUUGGAGCAGG
          Species: Human, Bovine, Pan troglodytes
          Product Type: TaqMan™ MicroRNA Assay
          Assay ID 462575_mat
          Size
          Availability Inventoried
          Catalog # 4427975
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000627, mir-1246

          Mature miRNA Sequence:

          AAUGGAUUUUUGGAGCAGG

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-1246 MIMAT0005898
          Bovine bta-miR-1246 MIMAT0024567
          Pan troglodytes ptr-miR-1246 MIMAT0007972

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-1246
          Stem-loop Accession # MI0006381
          Stem-loop Sequence
          UGUAUCCUUGAAUGGAUUUUUGGAGCAGGAGUGGACACCUGACCCAAAGGAAAUCAAUCCAUAGGCUAGCAAU
          Chromosome Location Chr. 2 - 177465708 - 177465780 [-] on Build 37
          Mature MicroRNA miRBase Accession #: MIMAT0005898
          miRBase ID: hsa-miR-1246
          Mature miRNA Sequence: AAUGGAUUUUUGGAGCAGG
          Chromosome Location: Chr. 2 - 177465708 - 177465780 [-] on Build 37

          Bovine

          Stem-loop ID bta-mir-1246
          Stem-loop Accession # MI0021112
          Stem-loop Sequence
          CAACAUAUUAAAUGGAUUUUUGGAGCAGGAAGUUGGAAUAGAGGCUUUCUCAGACAAAUACUUUACUGUGCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024567
          miRBase ID: bta-miR-1246
          Mature miRNA Sequence: AAUGGAUUUUUGGAGCAGG
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-1246
          Stem-loop Accession # MI0008452
          Stem-loop Sequence
          UGUAUCCUUGAAUGGAUUUUUGGAGCAGGAGUGGACACCUGACCCAAAGGAAAUCAAUCCAUAGGCUAGCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0007972
          miRBase ID: ptr-miR-1246
          Mature miRNA Sequence: AAUGGAUUUUUGGAGCAGG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03305724_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM13182
          Ambion® Pre-miR™ miRNA Precursor : PM13182
          mirVana® miRNA inhibitor : MH13182
          mirVana® miRNA mimic : MC13182

          Back To Top

          Related Products

          • TaqMan® Universal Master Mix II, with UNG
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® MicroRNA Reverse Transcription Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0