Search
Search

| Assay Name: | mmu-miR-130a* |
|---|---|
| miRBase Accession Number: | MI0000448 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GCUCUUUUCACAUUGUGCUACU |
| Species: | Human, Mouse, Rat, Chrysemys picta, Cricetulus griseus, Dasypus novemcinctus, Goat, Guinea pig, Monodelphis domestica, Ophiophagus hannah, Rabbit, Rhesus monkey |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000034, mir-130 |
Mature miRNA Sequence: | GCUCUUUUCACAUUGUGCUACU |
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-130a-5p | MIMAT0004593 | |
| Mouse | mmu-miR-130a-5p | MIMAT0016983 | |
| Rat | rno-miR-130a-5p | MIMAT0017121 | |
| Chrysemys picta | cpi-miR-130a-5p | MIMAT0037741 | |
| Cricetulus griseus | cgr-miR-130a-5p | MIMAT0023754 | |
| Dasypus novemcinctus | dno-miR-130a-5p | MIMAT0047627 | |
| View More Rows | |||
| Goat | chi-miR-130a-5p | MIMAT0035943 | |
| Guinea pig | cpo-miR-130a-5p | MIMAT0046980 | |
| Monodelphis domestica | mdo-miR-130a-5p | MIMAT0026664 | |
| Ophiophagus hannah | oha-miR-130c-5p | MIMAT0036691 | |
| Rabbit | ocu-miR-130a-5p | MIMAT0048217 | |
| Rhesus monkey | mml-miR-130a-5p | MIMAT0026566 | |