Search
Search

| Assay Name: | egr-miR-125 |
|---|---|
| miRBase Accession Number: | MI0017787 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UCCCUGAGACCCUAGAGUUGUC |
| Species: | Echinococcus granulosus, Echinococcus multilocularis |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000033, mir-10 |
Mature miRNA Sequence: | UCCCUGAGACCCUAGAGUUGUC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Echinococcus granulosus | egr-miR-125-5p | MIMAT0020243 |
| Echinococcus multilocularis | emu-miR-125-5p | MIMAT0020268 |