Search
Search

| Assay Name: | mmu-miR-5125 |
|---|---|
| miRBase Accession Number: | MI0018036 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UCUGCCUGGGAUUUCCUUGU |
| Species: | Mouse |
| Product Type: | TaqMan™ MicroRNA Assay |
Mature miRNA Sequence: | UCUGCCUGGGAUUUCCUUGU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-5125 | MIMAT0020635 |