Search
Search

| Assay Name: | smr-miR-965 |
|---|---|
| miRBase Accession Number: | MI0016441 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UAAGCGUAUGGCUUUUCCCCU |
| Species: | Strigamia maritima |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000708, mir-965 |
Mature miRNA Sequence: | UAAGCGUAUGGCUUUUCCCCU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Strigamia maritima | smr-miR-965 | MIMAT0018916 |