Search
Search

| Assay Name: | rno-miR-194* |
|---|---|
| miRBase Accession Number: | MI0000938 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CCAGUGGGGCUGCUGUUAUCU |
| Species: | Rat, Pig |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000055, mir-194 |
Mature miRNA Sequence: | CCAGUGGGGCUGCUGUUAUCU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-194-3p | MIMAT0017148 |
| Pig | ssc-miR-194a-3p | MIMAT0037465 |