Search
Search

| Assay Name: | pma-miR-10b |
|---|---|
| miRBase Accession Number: | MI0017009 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UACCCUGUAGGUACGACCUUGC |
| Species: | Petromyzon marinus |
| Product Type: | TaqMan™ MicroRNA Assay |
Mature miRNA Sequence: | UACCCUGUAGGUACGACCUUGC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Petromyzon marinus | pma-miR-10b-5p | MIMAT0019376 |