Search
Search

| Assay Name: | dme-miR-968-3p |
|---|---|
| miRBase Accession Number: | MI0005825 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | ACCUUUUGAUGUACUACUUUAA |
| Species: | Drosophila melanogaster |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000865, mir-968 |
Mature miRNA Sequence: | ACCUUUUGAUGUACUACUUUAA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Drosophila melanogaster | dme-miR-968-3p | MIMAT0020865 |