Search
Search

| Assay Name: | hsa-miR-3913-3p |
|---|---|
| miRBase Accession Number: | MI0016417 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AGACAUCAAGAUCAGUCCCAAA |
| Species: | Human |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0001134, mir-3913 |
Mature miRNA Sequence: | AGACAUCAAGAUCAGUCCCAAA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-3913-3p | MIMAT0019225 |