Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 465767_mat
          Assay Name: dre-miR-430a
          miRBase Accession Number: MI0023398
          miRBase Version: v22.1
          Chromosome Location: -
          Mature miRNA Sequence: UAAGUGCUAUUUGUUGGGGUAG
          Species: Cyprinus carpio, Hippoglossus hippoglossus, Zebrafish
          Product Type: TaqMan™ MicroRNA Assay
          Assay ID 465767_mat
          Size
          Availability Made To Order
          Catalog # 4440886
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000003, mir-430

          Mature miRNA Sequence:

          UAAGUGCUAUUUGUUGGGGUAG

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Cyprinus carpio ccr-miR-430 MIMAT0026298
          Hippoglossus hippoglossus hhi-miR-430a MIMAT0030341
          Zebrafish dre-miR-430a-3p MIMAT0001423

          Stem-loop Details



          Cyprinus carpio

          Stem-loop ID ccr-mir-430
          Stem-loop Accession # MI0023398
          Stem-loop Sequence
          CUAUCGGUACCCUCACAAUGGCACUGACUUGUCUGUUGUAUUUGGUAAGUGCUAUUUGUUGGGGUAGUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026298
          miRBase ID: ccr-miR-430
          Mature miRNA Sequence: UAAGUGCUAUUUGUUGGGGUAG
          Chromosome Location: NA

          Hippoglossus hippoglossus

          Stem-loop ID hhi-mir-430a-1
          Stem-loop Accession # MI0025420
          Stem-loop Sequence
          UUUAUCACCAAUAUUACCCUUGCACAAGCACCAACUUGAGUUUAGAAAACAGUAAGUGCUAUUUGUUGGGGUAGUAUUGAUGAUUUCAAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0030341
          miRBase ID: hhi-miR-430a
          Mature miRNA Sequence: UAAGUGCUAUUUGUUGGGGUAG
          Chromosome Location: NA

          Stem-loop ID hhi-mir-430a-2
          Stem-loop Accession # MI0025443
          Stem-loop Sequence
          AUCAAUUUAUCACCAAUAUUACCCUUGCACAAGCACCAACUUGAGUUUAGAAAACAGUAAGUGCUAUUUGUUGGGGUAGUAUUGAUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0030341
          miRBase ID: hhi-miR-430a
          Mature miRNA Sequence: UAAGUGCUAUUUGUUGGGGUAG
          Chromosome Location: NA

          Stem-loop ID hhi-mir-430a-3
          Stem-loop Accession # MI0025442
          Stem-loop Sequence
          ACUAGGUGGUCACCAGCUUUACCCUAACACAAGCAAAGACUUGCCUUCUGAAAGCAGUAAGUGCUAUUUGUUGGGGUAGUGUUGCUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0030341
          miRBase ID: hhi-miR-430a
          Mature miRNA Sequence: UAAGUGCUAUUUGUUGGGGUAG
          Chromosome Location: NA

          Zebrafish

          Stem-loop ID dre-mir-430a-1
          Stem-loop Accession # MI0001527
          Stem-loop Sequence
          GUCACUAUCGGUGCCCUCACAAAGGCACUGACUUGGAUGCUGCAUGUGGUAAGUGCUAUUUGUUGGGGUAGUUUCAAGUGAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001423
          miRBase ID: dre-miR-430a-3p
          Mature miRNA Sequence: UAAGUGCUAUUUGUUGGGGUAG
          Chromosome Location: NA

          Stem-loop ID dre-mir-430a-10
          Stem-loop Accession # MI0002119
          Stem-loop Sequence
          GUCACUAUCGGUACCCUCACAAAGGCACUGACUUGGAUGCUGUAAUUGGUAAGUGCUAUUUGUUGGGGUAGUUUCAAGUGAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001423
          miRBase ID: dre-miR-430a-3p
          Mature miRNA Sequence: UAAGUGCUAUUUGUUGGGGUAG
          Chromosome Location: NA

          Stem-loop ID dre-mir-430a-11
          Stem-loop Accession # MI0002120
          Stem-loop Sequence
          GUCUCUAUCGGUACCCUCACAAAAGCACUGACUUUGAUGCUGUAAUUGGUAAGUGCUAUUUGUUGGGGUAGUUUCAAGUGAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001423
          miRBase ID: dre-miR-430a-3p
          Mature miRNA Sequence: UAAGUGCUAUUUGUUGGGGUAG
          Chromosome Location: NA

          Stem-loop ID dre-mir-430a-12
          Stem-loop Accession # MI0002121
          Stem-loop Sequence
          GUCUCUAUCGGUACCCUCACAAAAGCACUGACUUUGAUGCUGUAUUUGGUAAGUGCUAUUUGUUGGGGUAGUUUCAAGUGAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001423
          miRBase ID: dre-miR-430a-3p
          Mature miRNA Sequence: UAAGUGCUAUUUGUUGGGGUAG
          Chromosome Location: NA

          Stem-loop ID dre-mir-430a-13
          Stem-loop Accession # MI0002122
          Stem-loop Sequence
          GUCUCUAUCGGUACCCUCACAAAAGCACUGACUUUGAUGCUGUAUUUGGUAAGUGCUAUUUGUUGGGGUAGUUUCAAGUGAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001423
          miRBase ID: dre-miR-430a-3p
          Mature miRNA Sequence: UAAGUGCUAUUUGUUGGGGUAG
          Chromosome Location: NA

          Stem-loop ID dre-mir-430a-14
          Stem-loop Accession # MI0002123
          Stem-loop Sequence
          GUCUCUAUCGGUACCCUCACAAAAGCACUGACUUUGAUGCUGUAUUUGGUAAGUGCUAUUUGUUGGGGUAGUUUCAAGUGAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001423
          miRBase ID: dre-miR-430a-3p
          Mature miRNA Sequence: UAAGUGCUAUUUGUUGGGGUAG
          Chromosome Location: NA

          Stem-loop ID dre-mir-430a-15
          Stem-loop Accession # MI0002124
          Stem-loop Sequence
          GUCUCUAUCGGUACCCUCACAAAAGCACUGACUUUGAUGCUGUAUUUGGUAAGUGCUAUUUGUUGGGGUAGUUUCAAGUGAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001423
          miRBase ID: dre-miR-430a-3p
          Mature miRNA Sequence: UAAGUGCUAUUUGUUGGGGUAG
          Chromosome Location: NA

          Stem-loop ID dre-mir-430a-16
          Stem-loop Accession # MI0002125
          Stem-loop Sequence
          GUCUCUAUCGGUACCCUCACAAAAGCACUGACUUUGAUGCUGUAUUUGGUAAGUGCUAUUUGUUGGGGUAGUUUUAAGUGAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001423
          miRBase ID: dre-miR-430a-3p
          Mature miRNA Sequence: UAAGUGCUAUUUGUUGGGGUAG
          Chromosome Location: NA

          Stem-loop ID dre-mir-430a-17
          Stem-loop Accession # MI0002126
          Stem-loop Sequence
          GUCUCUAUCGGUACCCUCACAAAAGCACUGACUUUGAUGCUGUAUUUGGUAAGUGCUAUUUGUUGGGGUAGUUUUAAGUGAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001423
          miRBase ID: dre-miR-430a-3p
          Mature miRNA Sequence: UAAGUGCUAUUUGUUGGGGUAG
          Chromosome Location: NA

          Stem-loop ID dre-mir-430a-18
          Stem-loop Accession # MI0002131
          Stem-loop Sequence
          GUCAGGCUUUCACAAGCCAGCCUCAAAGUUUGUUCUCAAACUUUAAGAAUCUAGUUGGUAAGUGCUAUUUGUUGGGGUAGUUUCAAGUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001423
          miRBase ID: dre-miR-430a-3p
          Mature miRNA Sequence: UAAGUGCUAUUUGUUGGGGUAG
          Chromosome Location: NA

          Stem-loop ID dre-mir-430a-2
          Stem-loop Accession # MI0002111
          Stem-loop Sequence
          GUCACUAUCGGUACCCUCACAAAGGCACUGACUUGGAUGCUGUAAUUGGUAAGUGCUAUUUGUUGGGGUAGUUUCAAGUGAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001423
          miRBase ID: dre-miR-430a-3p
          Mature miRNA Sequence: UAAGUGCUAUUUGUUGGGGUAG
          Chromosome Location: NA

          Stem-loop ID dre-mir-430a-3
          Stem-loop Accession # MI0002112
          Stem-loop Sequence
          GUCACUAUCGGUACCCUCACAAAGGCACUGACUUGGAUGCUGUAAUUGGUAAGUGCUAUUUGUUGGGGUAGUUUCAAGUGAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001423
          miRBase ID: dre-miR-430a-3p
          Mature miRNA Sequence: UAAGUGCUAUUUGUUGGGGUAG
          Chromosome Location: NA

          Stem-loop ID dre-mir-430a-4
          Stem-loop Accession # MI0002113
          Stem-loop Sequence
          GUCUCUAUCGGUACCCUCACAAAAGCACUGACUUUGAUGCUGUAUUUGGUAAGUGCUAUUUGUUGGGGUAGUUUCAAGUGAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001423
          miRBase ID: dre-miR-430a-3p
          Mature miRNA Sequence: UAAGUGCUAUUUGUUGGGGUAG
          Chromosome Location: NA

          Stem-loop ID dre-mir-430a-5
          Stem-loop Accession # MI0002114
          Stem-loop Sequence
          GUCACUAUCGGUACCCUCACAAAGGCACUGACUUGGAUGCUGUAAUUGGUAAGUGCUAUUUGUUGGGGUAGUUUCAAGUGAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001423
          miRBase ID: dre-miR-430a-3p
          Mature miRNA Sequence: UAAGUGCUAUUUGUUGGGGUAG
          Chromosome Location: NA

          Stem-loop ID dre-mir-430a-6
          Stem-loop Accession # MI0002115
          Stem-loop Sequence
          GUCACUAUCGGUACCCUCACAAAGGCACUGACUUGGAUGCUGUAAUUGGUAAGUGCUAUUUGUUGGGGUAGUUUCAAGUGAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001423
          miRBase ID: dre-miR-430a-3p
          Mature miRNA Sequence: UAAGUGCUAUUUGUUGGGGUAG
          Chromosome Location: NA

          Stem-loop ID dre-mir-430a-7
          Stem-loop Accession # MI0002116
          Stem-loop Sequence
          GUCACUAUCGGUACCCUCACAAAGGCACUGACUUGGAUGCUGUAAUUGGUAAGUGCUAUUUGUUGGGGUAGUUUCAAGUGAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001423
          miRBase ID: dre-miR-430a-3p
          Mature miRNA Sequence: UAAGUGCUAUUUGUUGGGGUAG
          Chromosome Location: NA

          Stem-loop ID dre-mir-430a-8
          Stem-loop Accession # MI0002117
          Stem-loop Sequence
          GUCACUAUCGGUACCCUCACAAAGGCACUGACUUGGAUGCUGUAAUUGGUAAGUGCUAUUUGUUGGGGUAGUUUCAAGUGAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001423
          miRBase ID: dre-miR-430a-3p
          Mature miRNA Sequence: UAAGUGCUAUUUGUUGGGGUAG
          Chromosome Location: NA

          Stem-loop ID dre-mir-430a-9
          Stem-loop Accession # MI0002118
          Stem-loop Sequence
          GUCACUAUCGGUACCCUCACAAAGGCACUGACUUGGAUGCUGUAAUUGGUAAGUGCUAUUUGUUGGGGUAGUUUCAAGUGAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001423
          miRBase ID: dre-miR-430a-3p
          Mature miRNA Sequence: UAAGUGCUAUUUGUUGGGGUAG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          mirVana® miRNA inhibitor : MH10393
          mirVana® miRNA mimic : MC10393


          miRBase Alias:

          Danio rerio: dre-miR-430a (v20)

          Back To Top

          Related Products

          • TaqMan® Universal Master Mix II, with UNG
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® MicroRNA Reverse Transcription Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline