Search
Search

| Assay Name: | hsa-miR-6767-5p |
|---|---|
| miRBase Accession Number: | MI0022612 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UCGCAGACAGGGACACAUGGAGA |
| Species: | Human |
| Product Type: | TaqMan™ MicroRNA Assay |
Mature miRNA Sequence: | UCGCAGACAGGGACACAUGGAGA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-6767-5p | MIMAT0027434 |