Search
Search

| Assay Name: | mml-miR-660-3p |
|---|---|
| miRBase Accession Number: | MI0007901 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | ACCUCCUGUGUGCAUGGAUUAC |
| Species: | Rhesus monkey |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000113, mir-188 |
Mature miRNA Sequence: | ACCUCCUGUGUGCAUGGAUUAC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rhesus monkey | mml-miR-660-3p | MIMAT0026907 |