Search
Search

| Assay Name: | mse-miR-750 |
|---|---|
| miRBase Accession Number: | MI0021006 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CCAGAUCUAUCUUUCCAGCUCA |
| Species: | Manduca sexta |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000796, mir-750 |
Mature miRNA Sequence: | CCAGAUCUAUCUUUCCAGCUCA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Manduca sexta | mse-miR-750 | MIMAT0024457 |