Search
Search

| Assay Name: | efu-miR-28 |
|---|---|
| miRBase Accession Number: | MI0028634 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AAGGAGCUCACAGUCUAUUGAGU |
| Species: | Eptesicus fuscus |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000057, mir-28 |
Mature miRNA Sequence: | AAGGAGCUCACAGUCUAUUGAGU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Eptesicus fuscus | efu-miR-28 | MIMAT0034949 |