Search
Search

| Assay Name: | chi-miR-660 |
|---|---|
| miRBase Accession Number: | MI0030828 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UACCCAUUGCAUAUCGGAGCUGU |
| Species: | Goat |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000113, mir-188 |
Mature miRNA Sequence: | UACCCAUUGCAUAUCGGAGCUGU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Goat | chi-miR-660 | MIMAT0036284 |