Search
Search

| Assay Name: | ath-miR8121 |
|---|---|
| miRBase Accession Number: | MI0026055 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AAAGUAUAAUGGUUUAGUGGUUUG |
| Species: | Arabidopsis |
| Product Type: | TaqMan™ MicroRNA Assay |
Mature miRNA Sequence: | AAAGUAUAAUGGUUUAGUGGUUUG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Arabidopsis | ath-miR8121 | MIMAT0032262 |