Search
Search

| Assay Name: | cgr-miR-7b |
|---|---|
| miRBase Accession Number: | MI0000730 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGGAAGACUUGUGAUUUUGUUGUU |
| Species: | Mouse, Alligator mississippiensis, Columba livia, Cricetulus griseus |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000022, mir-7 |
Mature miRNA Sequence: | UGGAAGACUUGUGAUUUUGUUGUU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-7b-5p | MIMAT0000678 |
| Alligator mississippiensis | ami-miR-7b-5p | MIMAT0038026 |
| Columba livia | cli-miR-7b-5p | MIMAT0038400 |
| Cricetulus griseus | cgr-miR-7b | MIMAT0024004 |