Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 477918_mir

          Specificity testing was performed using human anti-targets.

          Assay Name: hsa-miR-150-5p
          Stem-loop Accession Number: MI0000479
          miRBase Version: v22.1
          Chromosome Location: Chr.19: 49500785 - 49500868 [-] on Build GRCh38
          Mature miRNA Sequence: UCUCCCAACCCUUGUACCAGUG
          Species: Human, Mouse, Rat, Alligator mississippiensis, Anolis carolinensis, Callithrix jacchus, Cricetulus griseus, Dasypus novemcinctus, Dog, Goat, Gorilla gorilla, Guinea pig, Horse, Pan troglodytes, Pig, Pongo pygmaeus, Python bivittatus, Rhesus monkey, Sheep
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID 477918_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000197, mir-150

          Mature miRNA Sequence:

          UCUCCCAACCCUUGUACCAGUG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-150-5p MIMAT0000451
          Mouse mmu-miR-150-5p MIMAT0000160
          Rat rno-miR-150-5p MIMAT0000853
          Alligator mississippiensis ami-miR-150-5p MIMAT0038180
          Anolis carolinensis aca-miR-150-5p MIMAT0021784
          Callithrix jacchus cja-miR-150 MIMAT0039474
          View More Rows
          Cricetulus griseus cgr-miR-150 MIMAT0041150
          Dasypus novemcinctus dno-miR-150-5p MIMAT0047675
          Dog cfa-miR-150 MIMAT0006602
          Goat chi-miR-150 MIMAT0035980
          Gorilla gorilla ggo-miR-150 MIMAT0024217
          Guinea pig cpo-miR-150-5p MIMAT0047028
          Horse eca-miR-150 MIMAT0013011
          Pan troglodytes ptr-miR-150 MIMAT0008044
          Pig ssc-miR-150 MIMAT0025365
          Pongo pygmaeus ppy-miR-150 MIMAT0015774
          Python bivittatus pbv-miR-150-5p MIMAT0038968
          Rhesus monkey mml-miR-150-5p MIMAT0006211
          Sheep oar-miR-150 MIMAT0030034

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-150
          Stem-loop Accession # MI0000479
          Stem-loop Sequence
          CUCCCCAUGGCCCUGUCUCCCAACCCUUGUACCAGUGCUGGGCUCAGACCCUGGUACAGGCCUGGGGGACAGGGACCUGGGGAC
          Chromosome Location Chr. 19 - 49500785 - 49500868 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000451
          miRBase ID: hsa-miR-150-5p
          Mature miRNA Sequence: UCUCCCAACCCUUGUACCAGUG
          Chromosome Location: Chr. 19 - 49500785 - 49500868 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-150
          Stem-loop Accession # MI0000172
          Stem-loop Sequence
          CCCUGUCUCCCAACCCUUGUACCAGUGCUGUGCCUCAGACCCUGGUACAGGCCUGGGGGAUAGGG
          Chromosome Location Chr. 7 - 45121757 - 45121821 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000160
          miRBase ID: mmu-miR-150-5p
          Mature miRNA Sequence: UCUCCCAACCCUUGUACCAGUG
          Chromosome Location: Chr. 7 - 45121757 - 45121821 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-150
          Stem-loop Accession # MI0000920
          Stem-loop Sequence
          CUUCUCAAGGCCCUGUCUCCCAACCCUUGUACCAGUGCUGUGCCUCAGACCCUGGUACAGGCCUGGGGGACAGGGACUUGGGGAC
          Chromosome Location Chr. 1 - 101115974 - 101116058 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000853
          miRBase ID: rno-miR-150-5p
          Mature miRNA Sequence: UCUCCCAACCCUUGUACCAGUG
          Chromosome Location: Chr. 1 - 101115974 - 101116058 [+] on Build Rnor_6.0

          Alligator mississippiensis

          Stem-loop ID ami-mir-150
          Stem-loop Accession # MI0029756
          Stem-loop Sequence
          CCUUCUCUGCCCCCCACUCUCUCCCAACCCUUGUACCAGUGUCAUGUUACUGGAACCCUGGUACAGAGGAUGGAUGAGAAGGAGGCGUGGGACCCCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038180
          miRBase ID: ami-miR-150-5p
          Mature miRNA Sequence: UCUCCCAACCCUUGUACCAGUG
          Chromosome Location: NA

          Anolis carolinensis

          Stem-loop ID aca-mir-150
          Stem-loop Accession # MI0018761
          Stem-loop Sequence
          AAACUCUCUUUCCCUCUCUCCCAACCCUUGUACCAGUGCCAACUGUAGUCCUCACUGGUACAGAGGAUGGAAGGGUGGGCCUGCAGAGAUGAGACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021784
          miRBase ID: aca-miR-150-5p
          Mature miRNA Sequence: UCUCCCAACCCUUGUACCAGUG
          Chromosome Location: NA

          Callithrix jacchus

          Stem-loop ID cja-mir-150
          Stem-loop Accession # MI0031998
          Stem-loop Sequence
          UCCCCAUAGCCCUGUCUCCCAACCCUUGUACCAGUGCUGGGCUCAGACCCUGGUACAGGCCUGGGGGGCAGGGACCUGGGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039474
          miRBase ID: cja-miR-150
          Mature miRNA Sequence: UCUCCCAACCCUUGUACCAGUG
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-150
          Stem-loop Accession # MI0033079
          Stem-loop Sequence
          CCCUGUCUCCCAACCCUUGUACCAGUGCUGCACUUCAGACCCUGGUACAGGCCUGGGGGGCAGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0041150
          miRBase ID: cgr-miR-150
          Mature miRNA Sequence: UCUCCCAACCCUUGUACCAGUG
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-150
          Stem-loop Accession # MI0039000
          Stem-loop Sequence
          UCUCCCAACCCUUGUACCAGUGCUGGCCUCAGACCCUGGUACAGGCCUGGGGGGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047675
          miRBase ID: dno-miR-150-5p
          Mature miRNA Sequence: UCUCCCAACCCUUGUACCAGUG
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-150
          Stem-loop Accession # MI0007998
          Stem-loop Sequence
          UCUCCCAACCCUUGUACCAGUGCUGUGCCUCAGUCCCUGGUACAGGCAUGGGGGGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006602
          miRBase ID: cfa-miR-150
          Mature miRNA Sequence: UCUCCCAACCCUUGUACCAGUG
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-150
          Stem-loop Accession # MI0030638
          Stem-loop Sequence
          UCUCUUCUCAUGGCCCUGUCUCCCAACCCUUGUACCAGUGUGUGUCUUGGUCCCUGGUACAGGUACGGGGAGGCAGGGACCUGGGGAUCCCAGCAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0035980
          miRBase ID: chi-miR-150
          Mature miRNA Sequence: UCUCCCAACCCUUGUACCAGUG
          Chromosome Location: NA

          Gorilla gorilla

          Stem-loop ID ggo-mir-150
          Stem-loop Accession # MI0020764
          Stem-loop Sequence
          CUCCUCUCCCCAUGGCCCUGUCUCCCAACCCUUGUACCAGUGCUGGGCUCAGACCCUGGUACAGGCCUGGGGGACAGGGACCUGGGGACCCCGGCACCGGCAGGCCCCAAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024217
          miRBase ID: ggo-miR-150
          Mature miRNA Sequence: UCUCCCAACCCUUGUACCAGUG
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-150
          Stem-loop Accession # MI0038659
          Stem-loop Sequence
          UCUCCCAACCCUUGUACCAGUGCUGUGCCUCAGACGCUGGUACAGGCCUGGGGGGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047028
          miRBase ID: cpo-miR-150-5p
          Mature miRNA Sequence: UCUCCCAACCCUUGUACCAGUG
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-150
          Stem-loop Accession # MI0012762
          Stem-loop Sequence
          CCCUGUCUCCCAACCCUUGUACCAGUGCUGUGCCUCGGUCCCUGGUACAGGCAUGGGGGGCAGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013011
          miRBase ID: eca-miR-150
          Mature miRNA Sequence: UCUCCCAACCCUUGUACCAGUG
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-150
          Stem-loop Accession # MI0008550
          Stem-loop Sequence
          CUCCCCAUGGCCCUGUCUCCCAACCCUUGUACCAGUGCUGGGCUCAGACCCUGGUACAGGCCUGGGGGACAGGGACCUGGGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008044
          miRBase ID: ptr-miR-150
          Mature miRNA Sequence: UCUCCCAACCCUUGUACCAGUG
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-150-1
          Stem-loop Accession # MI0022133
          Stem-loop Sequence
          CUCGGCCCUAUCUCCCAACCCUUGUACCAGUGCGUGCCUCAGUCCCUGGUACAGGAAUGGGGGGCAGGGACCUGGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0025365
          miRBase ID: ssc-miR-150
          Mature miRNA Sequence: UCUCCCAACCCUUGUACCAGUG
          Chromosome Location: NA

          Stem-loop ID ssc-mir-150-2
          Stem-loop Accession # MI0022134
          Stem-loop Sequence
          CUCGGCCCUAUCUCCCAACCCUUGUACCAGUGCGUGCCUCAGUCCCUGGUACAGGAAUGGGGGGCAGGGACCUGGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0025365
          miRBase ID: ssc-miR-150
          Mature miRNA Sequence: UCUCCCAACCCUUGUACCAGUG
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-150
          Stem-loop Accession # MI0014840
          Stem-loop Sequence
          CUCCCCAUGGCCCUGUCUCCCAACCCUUGUACCAGUGCUGGGCUCAGACCCUGGUACAGGCCUGGGGGACAGGGACCUGGGGAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015774
          miRBase ID: ppy-miR-150
          Mature miRNA Sequence: UCUCCCAACCCUUGUACCAGUG
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-mir-150
          Stem-loop Accession # MI0030249
          Stem-loop Sequence
          ACUCUUCUCCUUUCUCUCCCAACCCUUGUACCAGUGUCAGCUGUUGCACCUACUGGUACAGAGGAUGGAAGGGAGAGUGGUUCAGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038968
          miRBase ID: pbv-miR-150-5p
          Mature miRNA Sequence: UCUCCCAACCCUUGUACCAGUG
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-150
          Stem-loop Accession # MI0007641
          Stem-loop Sequence
          CUCCCCAUGGCCCUGUCUCCCAACCCUUGUACCAGUGCUGGGCUCAGACCCUGGUACAGGCCUGGGGGACAGGGACCUGGGGAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006211
          miRBase ID: mml-miR-150-5p
          Mature miRNA Sequence: UCUCCCAACCCUUGUACCAGUG
          Chromosome Location: NA

          Sheep

          Stem-loop ID oar-mir-150
          Stem-loop Accession # MI0025255
          Stem-loop Sequence
          UCUCUCCUCACGGCCCUGUCUCCCAACCCUUGUACCAGUGUGUGUCUCGGUCCCUGGUACAGGUACGGGGAGGCAGGGACCUGGGGAUCCCAGCAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0030034
          miRBase ID: oar-miR-150
          Mature miRNA Sequence: UCUCCCAACCCUUGUACCAGUG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03466044_pri
          TaqMan™ Pri-miRNA Assay : Hs03303271_pri
          TaqMan™ Pri-miRNA Assay : Mm03306366_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10070
          Ambion® Pre-miR™ miRNA Precursor : PM10070
          TaqMan™ MicroRNA Assay : 000473
          mirVana® miRNA inhibitor : MH10070
          mirVana® miRNA mimic : MC10070
          TaqMan™ Advanced miRNA Assay : rno480947_mir
          TaqMan™ Advanced miRNA Assay : mmu480947_mir
          MIRNA : TaqMan® Advanced miRNA Human A and B 96-well Plates, Fast
          MIRNA : TaqMan® Advanced miRNA Human Serum/Plasma Card
          FGMIR : QS TaqmanTM OpenArray Human Advanced miRNA Panel
          MIRNA : TaqMan® Advanced miRNA Human Serum/Plasma 96-well Plates, Standard
          FGMIR : TaqManTM Advanced miRNA Human Serum/Plasma 96-well Plate 1
          FGMIR : TaqManTM Advanced miRNA Human A Card
          FGMIR : TaqManTM Advanced miRNA Human A 96-well Plate 2
          MIRNA : TaqMan® Advanced miRNA Human A Card
          MIRNA : TaqMan® Advanced miRNA Human A and B 96-well Plates, Standard
          MIRNA : TaqMan® OpenArray® Human Advanced MicroRNA Panel
          MIRNA : TaqMan® Advanced miRNA Human A and B Cards
          MIRNA : TaqMan® Advanced miRNA Human Serum/Plasma 96-well Plates, Fast
          FGMIR : TaqManTM Advanced miRNA Human Serum/Plasma Card
          FGMIR : TaqManTM Advanced miRNA Human Serum/Plasma 96-well Plate 1
          MIRNA : TaqMan® Advanced miRNA Human A 96-well Plates, Standard
          FGMIR : TaqManTM Advanced miRNA Human A 96-well Plate 2
          MIRNA : TaqMan® Advanced miRNA Human A 96-well Plates, Fast

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline