Search
Search

| Assay Name: | hsa-miR-31-5p |
|---|---|
| Stem-loop Accession Number: | MI0000089 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AGGCAAGAUGCUGGCAUAGCU |
| Species: | Human, Bovine, Horse, Pteropus alecto |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000064, mir-31 |
Mature miRNA Sequence: | AGGCAAGAUGCUGGCAUAGCU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-31-5p | MIMAT0000089 |
| Bovine | bta-miR-31 | MIMAT0003548 |
| Horse | eca-miR-31 | MIMAT0013115 |
| Pteropus alecto | pal-miR-31-5p | MIMAT0040136 |