Search
Search

| Assay Name: | hsa-miR-342-5p |
|---|---|
| Stem-loop Accession Number: | MI0000805 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AGGGGUGCUAUCUGUGAUUGA |
| Species: | Human, Pongo pygmaeus, Pteropus alecto, Rhesus monkey |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000190, mir-342 |
Mature miRNA Sequence: | AGGGGUGCUAUCUGUGAUUGA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-342-5p | MIMAT0004694 |
| Pongo pygmaeus | ppy-miR-342-5p | MIMAT0015844 |
| Pteropus alecto | pal-miR-342-5p | MIMAT0040085 |
| Rhesus monkey | mml-miR-342-5p | MIMAT0006282 |