Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 478090_mir

          Specificity testing was performed using human anti-targets.

          Assay Name: hsa-miR-423-5p
          Stem-loop Accession Number: MI0001445
          miRBase Version: v22.1
          Chromosome Location: Chr.17: 30117079 - 30117172 [+] on Build GRCh38
          Mature miRNA Sequence: UGAGGGGCAGAGAGCGAGACUUU
          Species: Human, Mouse, Bovine, Callithrix jacchus, Cricetulus griseus, Dasypus novemcinctus, Dog, Goat, Guinea pig, Horse, Papio hamadryas, Pig, Pongo pygmaeus, Pteropus alecto, Rabbit, Rhesus monkey
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID 478090_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000329, mir-423

          Mature miRNA Sequence:

          UGAGGGGCAGAGAGCGAGACUUU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-423-5p MIMAT0004748
          Mouse mmu-miR-423-5p MIMAT0004825
          Bovine bta-miR-423-5p MIMAT0012537
          Callithrix jacchus cja-miR-423-5p MIMAT0039483
          Cricetulus griseus cgr-miR-423-5p MIMAT0023956
          Dasypus novemcinctus dno-miR-423-5p MIMAT0047880
          View More Rows
          Dog cfa-miR-423a MIMAT0006742
          Goat chi-miR-423-5p MIMAT0036220
          Guinea pig cpo-miR-423-5p MIMAT0047241
          Horse eca-miR-423-5p MIMAT0013039
          Papio hamadryas pha-miR-423 MIMAT0049512
          Pig ssc-miR-423-5p MIMAT0013880
          Pongo pygmaeus ppy-miR-423-5p MIMAT0015882
          Pteropus alecto pal-miR-423-5p MIMAT0039830
          Rabbit ocu-miR-423-5p MIMAT0048441
          Rhesus monkey mml-miR-423-5p MIMAT0006321

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-423
          Stem-loop Accession # MI0001445
          Stem-loop Sequence
          AUAAAGGAAGUUAGGCUGAGGGGCAGAGAGCGAGACUUUUCUAUUUUCCAAAAGCUCGGUCUGAGGCCCCUCAGUCUUGCUUCCUAACCCGCGC
          Chromosome Location Chr. 17 - 30117079 - 30117172 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004748
          miRBase ID: hsa-miR-423-5p
          Mature miRNA Sequence: UGAGGGGCAGAGAGCGAGACUUU
          Chromosome Location: Chr. 17 - 30117079 - 30117172 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-423
          Stem-loop Accession # MI0004637
          Stem-loop Sequence
          ACUUGUGAGGAAAUAAAGGAAGUUAGGCUGAGGGGCAGAGAGCGAGACUUUUCUAUUUUCCAAAAGCUCGGUCUGAGGCCCCUCAGUCUUGCUUCCUACCCCGCGCUUG
          Chromosome Location Chr. 11 - 77078064 - 77078172 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004825
          miRBase ID: mmu-miR-423-5p
          Mature miRNA Sequence: UGAGGGGCAGAGAGCGAGACUUU
          Chromosome Location: Chr. 11 - 77078064 - 77078172 [-] on Build GRCm38

          Bovine

          Stem-loop ID bta-mir-423
          Stem-loop Accession # MI0005046
          Stem-loop Sequence
          AUAAAGGAAGUUAGGCUGAGGGGCAGAGAGCGAGACUUUUCUAUUUUCCAAAAGCUCGGUCUGAGGCCCCUCAGUCUUGCUUCCUACCCCGCGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012537
          miRBase ID: bta-miR-423-5p
          Mature miRNA Sequence: UGAGGGGCAGAGAGCGAGACUUU
          Chromosome Location: NA

          Callithrix jacchus

          Stem-loop ID cja-mir-423
          Stem-loop Accession # MI0032008
          Stem-loop Sequence
          CCGUACAUUUUCCAGGAUGGAAGCCCGAAGUUUGAGGGAGAAACUUGUGAGGAAAUAAAGGAAGUUAGGCUGAGGGGCAGAGAGCGAGACUUUUCUAUUUUCCAAAAGCUCGGUCUGAGGCCCCUCAGUCUUGCUUCCUACCCCGCGCUUGAGUUUCUCCCCGCUUGGAUGCUCUCAGGGGCAGUGUGAAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039483
          miRBase ID: cja-miR-423-5p
          Mature miRNA Sequence: UGAGGGGCAGAGAGCGAGACUUU
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-423
          Stem-loop Accession # MI0020523
          Stem-loop Sequence
          GAAGUUAGGCUGAGGGGCAGAGAGCGAGACUUUUCUAUUUUCCAAAAGCUCGGUCUGAGGCCCCUCAGUCUUGCUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023956
          miRBase ID: cgr-miR-423-5p
          Mature miRNA Sequence: UGAGGGGCAGAGAGCGAGACUUU
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-423
          Stem-loop Accession # MI0039105
          Stem-loop Sequence
          UGAGGGGCAGAGAGCGAGACUUUUCUAUUUUCCAAAAGCUCGGUCUGAGGCCCCUCAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047880
          miRBase ID: dno-miR-423-5p
          Mature miRNA Sequence: UGAGGGGCAGAGAGCGAGACUUU
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-423a
          Stem-loop Accession # MI0008166
          Stem-loop Sequence
          UGAGGGGCAGAGAGCGAGACUUUUCUAUUUUCCAAAAGCUCGGUCUGAGGCCCCUCAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006742
          miRBase ID: cfa-miR-423a
          Mature miRNA Sequence: UGAGGGGCAGAGAGCGAGACUUU
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-423
          Stem-loop Accession # MI0030788
          Stem-loop Sequence
          ACUUGUGAGGAAAUAAAGGAAGUUAGGCUGAGGGGCAGAGAGCGAGACUUUUCUAUUUUCCAAAAGCUCGGUCUGAGGCCCCUCAGUCUUGCUUCCUACCCCGCGCUUGAGUUUCUCCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036220
          miRBase ID: chi-miR-423-5p
          Mature miRNA Sequence: UGAGGGGCAGAGAGCGAGACUUU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-423
          Stem-loop Accession # MI0038769
          Stem-loop Sequence
          UGAGGGGCAGAGAGCGAGACUUUUCUAUUUUCCAAAAGCUCGGUCUGAGGCCCCUCAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047241
          miRBase ID: cpo-miR-423-5p
          Mature miRNA Sequence: UGAGGGGCAGAGAGCGAGACUUU
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-423
          Stem-loop Accession # MI0012784
          Stem-loop Sequence
          AGGCUGAGGGGCAGAGAGCGAGACUUUUCUAUUUUCCAAAAGCUCGGUCUGAGGCCCCUCAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013039
          miRBase ID: eca-miR-423-5p
          Mature miRNA Sequence: UGAGGGGCAGAGAGCGAGACUUU
          Chromosome Location: NA

          Papio hamadryas

          Stem-loop ID pha-mir-423
          Stem-loop Accession # MI0040277
          Stem-loop Sequence
          UGAGGGGCAGAGAGCGAGACUUUUCUAUUUUCCAAAAGCUCGGUCUGAGGCCCCUCAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049512
          miRBase ID: pha-miR-423
          Mature miRNA Sequence: UGAGGGGCAGAGAGCGAGACUUU
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-423
          Stem-loop Accession # MI0013099
          Stem-loop Sequence
          AAGUUAGGCUGAGGGGCAGAGAGCGAGACUUUUCUAUUUUCCAAAAGCUCGGUCUGAGGCCCCUCAGUCUUGCUUCCUAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013880
          miRBase ID: ssc-miR-423-5p
          Mature miRNA Sequence: UGAGGGGCAGAGAGCGAGACUUU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-423
          Stem-loop Accession # MI0014939
          Stem-loop Sequence
          AGGAAGUUAGGCUGAGGGGCAGAGAGCGAGACUUUUCUAUUUUCCAAAAGCUCGGUCUGAGGCCCCUCAGUCUUGCUUCCUACCCCGCGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015882
          miRBase ID: ppy-miR-423-5p
          Mature miRNA Sequence: UGAGGGGCAGAGAGCGAGACUUU
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-423
          Stem-loop Accession # MI0032351
          Stem-loop Sequence
          UGAGGGGCAGAGAGCGAGACUUUUCUAUUUUCCAAAAGCUCGGUCUGAGGCCCCUCAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039830
          miRBase ID: pal-miR-423-5p
          Mature miRNA Sequence: UGAGGGGCAGAGAGCGAGACUUU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-423
          Stem-loop Accession # MI0039402
          Stem-loop Sequence
          UGAGGGGCAGAGAGCGAGACUUUUCUAUUUUCCAAAAGCUCGGUCUGAGGCCCCUCAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048441
          miRBase ID: ocu-miR-423-5p
          Mature miRNA Sequence: UGAGGGGCAGAGAGCGAGACUUU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-423
          Stem-loop Accession # MI0007739
          Stem-loop Sequence
          AUAAAGGAAGUUAGGCUGAGGGGCAGAGAGCGAGACUUUUCUAUUUUCCAAAAGCUCGGUCUGAGGCCCCUCAGUCUUGCUUCCUACCCCGCGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006321
          miRBase ID: mml-miR-423-5p
          Mature miRNA Sequence: UGAGGGGCAGAGAGCGAGACUUU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03307970_pri
          TaqMan™ Pri-miRNA Assay : Hs03303690_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM11164
          Ambion® Pre-miR™ miRNA Precursor : PM11164
          TaqMan™ MicroRNA Assay : 002340
          mirVana® miRNA inhibitor : MH11164
          mirVana® miRNA mimic : MC11164
          TaqMan™ Advanced miRNA Assay : mmu481834_mir
          MIRNA : TaqMan® Advanced miRNA Human Serum/Plasma 96-well Plates, Standard
          MIRNA : TaqMan® Advanced miRNA Human A and B 96-well Plates, Fast
          FGMIR : TaqManTM Advanced miRNA Human Serum/Plasma 96-well Plate 2
          FGMIR : TaqManTM Advanced miRNA Human Serum/Plasma Card
          MIRNA : TaqMan® Advanced miRNA Human Endogenous Controls 96-well Plate, Fast
          MIRNA : TaqMan® Advanced miRNA Human A Card
          FGMIR : TaqManTM Advanced miRNA Human A 96-well Plate 3
          MIRNA : TaqMan® Advanced miRNA Human A 96-well Plates, Fast
          FGMIR : TaqManTM Advanced miRNA Human A 96-well Plate 3
          FGMIR : TaqManTM Advanced miRNA Human Endogenous Controls 96-well Plate
          MIRNA : TaqMan® Advanced miRNA Human Serum/Plasma 96-well Plates, Fast
          MIRNA : TaqMan® Advanced miRNA Human Serum/Plasma Card
          FGMIR : TaqManTM Advanced miRNA Human Serum/Plasma 96-well Plate 2
          FGMIR : QS TaqmanTM OpenArray Human Advanced miRNA Panel
          MIRNA : TaqMan® Advanced miRNA Human A 96-well Plates, Standard
          MIRNA : TaqMan® Advanced miRNA Human A and B Cards
          MIRNA : TaqMan® OpenArray® Human Advanced MicroRNA Panel
          FGMIR : TaqManTM Advanced miRNA Human Endogenous Controls 96-well Plate
          FGMIR : TaqManTM Advanced miRNA Human A Card
          FGMIR : TaqManTM Advanced miRNA Human Endogenous Controls Card
          MIRNA : TaqMan® Advanced miRNA Human A and B 96-well Plates, Standard
          MIRNA : TaqMan® Advanced miRNA Human Endogenous Controls 96-well Plate, Standard
          MIRNA : TaqMan® Advanced miRNA Human Endogenous Controls Card

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline