Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 478113_mir

          Specificity testing was performed using human anti-targets.

          Assay Name: hsa-miR-455-5p
          Stem-loop Accession Number: MI0003513
          miRBase Version: v22.1
          Chromosome Location: Chr.9: 114209434 - 114209529 [+] on Build GRCh38
          Mature miRNA Sequence: UAUGUGCCUUUGGACUACAUCG
          Species: Human, Mouse, Rat, Callithrix jacchus, Cricetulus griseus, Dasypus novemcinctus, Dog, Goat, Guinea pig, Nomascus leucogenys, Pig, Pongo pygmaeus, Rabbit, Rhesus monkey
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID 478113_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000129, mir-455

          Mature miRNA Sequence:

          UAUGUGCCUUUGGACUACAUCG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-455-5p MIMAT0003150
          Mouse mmu-miR-455-5p MIMAT0003485
          Rat rno-miR-455-5p MIMAT0005316
          Callithrix jacchus cja-miR-455 MIMAT0039451
          Cricetulus griseus cgr-miR-455-5p MIMAT0023964
          Dasypus novemcinctus dno-miR-455-5p MIMAT0047906
          View More Rows
          Dog cfa-miR-455 MIMAT0006603
          Goat chi-miR-455-5p MIMAT0036241
          Guinea pig cpo-miR-455-5p MIMAT0047271
          Nomascus leucogenys nle-miR-455 MIMAT0049432
          Pig ssc-miR-455-5p MIMAT0022957
          Pongo pygmaeus ppy-miR-455-5p MIMAT0015899
          Rabbit ocu-miR-455-5p MIMAT0048465
          Rhesus monkey mml-miR-455-5p MIMAT0006339

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-455
          Stem-loop Accession # MI0003513
          Stem-loop Sequence
          UCCCUGGCGUGAGGGUAUGUGCCUUUGGACUACAUCGUGGAAGCCAGCACCAUGCAGUCCAUGGGCAUAUACACUUGCCUCAAGGCCUAUGUCAUC
          Chromosome Location Chr. 9 - 114209434 - 114209529 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0003150
          miRBase ID: hsa-miR-455-5p
          Mature miRNA Sequence: UAUGUGCCUUUGGACUACAUCG
          Chromosome Location: Chr. 9 - 114209434 - 114209529 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-455
          Stem-loop Accession # MI0004679
          Stem-loop Sequence
          CUCCCUGGUGUGAGCGUAUGUGCCUUUGGACUACAUCGUGAACGCAGCACCAUGCAGUCCACGGGCAUAUACACUUGCCUCA
          Chromosome Location Chr. 4 - 63256851 - 63256932 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0003485
          miRBase ID: mmu-miR-455-5p
          Mature miRNA Sequence: UAUGUGCCUUUGGACUACAUCG
          Chromosome Location: Chr. 4 - 63256851 - 63256932 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-455
          Stem-loop Accession # MI0006148
          Stem-loop Sequence
          GGUGUGAGCGUAUGUGCCUUUGGACUACAUCGUGGACGCAGCACCAUGCAGUCCACGGGCAUAUACACUUGCCUCAAG
          Chromosome Location Chr. 5 - 79097208 - 79097285 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0005316
          miRBase ID: rno-miR-455-5p
          Mature miRNA Sequence: UAUGUGCCUUUGGACUACAUCG
          Chromosome Location: Chr. 5 - 79097208 - 79097285 [+] on Build Rnor_6.0

          Callithrix jacchus

          Stem-loop ID cja-mir-455
          Stem-loop Accession # MI0031974
          Stem-loop Sequence
          GUAUGUGCCUUUGGACUACAUCGUGGAAGCCAGCACCAUGCAGUCCAUGGGCAUAUAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039451
          miRBase ID: cja-miR-455
          Mature miRNA Sequence: UAUGUGCCUUUGGACUACAUCG
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-455
          Stem-loop Accession # MI0020528
          Stem-loop Sequence
          GGUGUGAGCGUAUGUGCCUUUGGACUACAUCGUGGACGCAGCACCAUGCAGUCCACGGGCAUAUACACUUGCCUCAAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023964
          miRBase ID: cgr-miR-455-5p
          Mature miRNA Sequence: UAUGUGCCUUUGGACUACAUCG
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-455
          Stem-loop Accession # MI0039119
          Stem-loop Sequence
          UAUGUGCCUUUGGACUACAUCGUGGAAGCCAGCACCAUGCAGUCCGUGGGCAUAUACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047906
          miRBase ID: dno-miR-455-5p
          Mature miRNA Sequence: UAUGUGCCUUUGGACUACAUCG
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-455
          Stem-loop Accession # MI0007999
          Stem-loop Sequence
          UAUGUGCCUUUGGACUACAUCGUGGAAGCCAGCACCAUGCAGUCCAUGGGCAUAUACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006603
          miRBase ID: cfa-miR-455
          Mature miRNA Sequence: UAUGUGCCUUUGGACUACAUCG
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-455
          Stem-loop Accession # MI0030800
          Stem-loop Sequence
          GCGAGUGCCCGCGCCCUUCCCUGGCGUGAGGGUAUGUGCCUUUGGACUACAUCGUGGAAGCCAGCACCAUGCAGUCCAUGGGCAUAUACACUUGCCUCAAGGCCUAUGUCAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036241
          miRBase ID: chi-miR-455-5p
          Mature miRNA Sequence: UAUGUGCCUUUGGACUACAUCG
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-455
          Stem-loop Accession # MI0038785
          Stem-loop Sequence
          UAUGUGCCUUUGGACUACAUCGUGGAAGCCAGCACCAUGCAGUCCAUGGGCAUAUACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047271
          miRBase ID: cpo-miR-455-5p
          Mature miRNA Sequence: UAUGUGCCUUUGGACUACAUCG
          Chromosome Location: NA

          Nomascus leucogenys

          Stem-loop ID nle-mir-455
          Stem-loop Accession # MI0040177
          Stem-loop Sequence
          UAUGUGCCUUUGGACUACAUCGUGGAAGCCAGCACCAUGCAGUCCAUGGGCAUAUACACU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049432
          miRBase ID: nle-miR-455
          Mature miRNA Sequence: UAUGUGCCUUUGGACUACAUCG
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-455
          Stem-loop Accession # MI0013170
          Stem-loop Sequence
          UGGCGUGAGGGUAUGUGCCUUUGGACUACAUCGUGGAAGCCAGCACCAUGCAGUCCAUGGGCAUAUACACUUGCCUCAAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0022957
          miRBase ID: ssc-miR-455-5p
          Mature miRNA Sequence: UAUGUGCCUUUGGACUACAUCG
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-455
          Stem-loop Accession # MI0014955
          Stem-loop Sequence
          UCCCUGGCGUGAGGGUAUGUGCCUUUGGACUACAUCGUGGAAGCCAGCACCAUGCAGUCCAUGGGCAUAUACACUUGCCUCAAGGCCUAUGUCAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015899
          miRBase ID: ppy-miR-455-5p
          Mature miRNA Sequence: UAUGUGCCUUUGGACUACAUCG
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-455
          Stem-loop Accession # MI0039415
          Stem-loop Sequence
          UAUGUGCCUUUGGACUACAUCGUGGAAGCCAGCACCAUGCAGUCCAUGGGCAUAUACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048465
          miRBase ID: ocu-miR-455-5p
          Mature miRNA Sequence: UAUGUGCCUUUGGACUACAUCG
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-455
          Stem-loop Accession # MI0007756
          Stem-loop Sequence
          UCCCUGGCGUGAGGGUAUGUGCCUUUGGACUACAUCGUGGAAGCCAGCACCAUGCAGUCCAUGGGCAUAUACACUUGCCUCAAGGCCUAUGUCAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006339
          miRBase ID: mml-miR-455-5p
          Mature miRNA Sequence: UAUGUGCCUUUGGACUACAUCG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03308178_pri
          TaqMan™ Pri-miRNA Assay : Rn03464700_pri
          TaqMan™ Pri-miRNA Assay : Hs03294041_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10529
          Ambion® Pre-miR™ miRNA Precursor : PM10529
          TaqMan™ MicroRNA Assay : 001280
          mirVana® miRNA inhibitor : MH10529
          mirVana® miRNA mimic : MC10529
          TaqMan™ Advanced miRNA Assay : mmu481173_mir
          TaqMan™ Advanced miRNA Assay : rno481173_mir
          MIRNA : TaqMan® Advanced miRNA Human A Card
          MIRNA : TaqMan® Advanced miRNA Human A 96-well Plates, Fast
          MIRNA : TaqMan® OpenArray® Human Advanced MicroRNA Panel
          MIRNA : TaqMan® Advanced miRNA Human A and B 96-well Plates, Standard
          FGMIR : TaqManTM Advanced miRNA Human A 96-well Plate 3
          FGMIR : TaqManTM Advanced miRNA Human A Card
          FGMIR : QS TaqmanTM OpenArray Human Advanced miRNA Panel
          MIRNA : TaqMan® Advanced miRNA Human A 96-well Plates, Standard
          MIRNA : TaqMan® Advanced miRNA Human A and B 96-well Plates, Fast
          FGMIR : TaqManTM Advanced miRNA Human A 96-well Plate 3
          MIRNA : TaqMan® Advanced miRNA Human A and B Cards

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline