Search
Search

| Assay Name: | cel-miR-238-3p |
|---|---|
| Stem-loop Accession Number: | MI0000313 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UUUGUACUCCGAUGCCAUUCAGA |
| Species: | Caenorhabditis elegans, Caenorhabditis brenneri |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000862, mir-238 |
Mature miRNA Sequence: | UUUGUACUCCGAUGCCAUUCAGA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Caenorhabditis elegans | cel-miR-238-3p | MIMAT0000293 |
| Caenorhabditis brenneri | cbn-miR-238 | MIMAT0029726 |