Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 478705_mir

          Specificity testing was performed using human anti-targets.

          Assay Name: hsa-miR-132-5p
          Stem-loop Accession Number: MI0000449
          miRBase Version: v22.1
          Chromosome Location: Chr.17: 2049908 - 2050008 [-] on Build GRCh38
          Mature miRNA Sequence: ACCGUGGCUUUCGAUUGUUACU
          Species: Human, Rat, Cricetulus griseus, Dasypus novemcinctus, Guinea pig, Monodelphis domestica, Pteropus alecto, Rhesus monkey
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID 478705_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000065, mir-132

          Mature miRNA Sequence:

          ACCGUGGCUUUCGAUUGUUACU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-132-5p MIMAT0004594
          Rat rno-miR-132-5p MIMAT0017123
          Cricetulus griseus cgr-miR-132-5p MIMAT0023758
          Dasypus novemcinctus dno-miR-132-5p MIMAT0047631
          Guinea pig cpo-miR-132-5p MIMAT0046984
          Monodelphis domestica mdo-miR-132-5p MIMAT0026693
          View More Rows
          Pteropus alecto pal-miR-132-5p MIMAT0039991
          Rhesus monkey mml-miR-132-5p MIMAT0026816

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-132
          Stem-loop Accession # MI0000449
          Stem-loop Sequence
          CCGCCCCCGCGUCUCCAGGGCAACCGUGGCUUUCGAUUGUUACUGUGGGAACUGGAGGUAACAGUCUACAGCCAUGGUCGCCCCGCAGCACGCCCACGCGC
          Chromosome Location Chr. 17 - 2049908 - 2050008 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004594
          miRBase ID: hsa-miR-132-5p
          Mature miRNA Sequence: ACCGUGGCUUUCGAUUGUUACU
          Chromosome Location: Chr. 17 - 2049908 - 2050008 [-] on Build GRCh38

          Rat

          Stem-loop ID rno-mir-132
          Stem-loop Accession # MI0000905
          Stem-loop Sequence
          CCGCCCCCGCGUCUCCAGGGCAACCGUGGCUUUCGAUUGUUACUGUGGGAACCGGAGGUAACAGUCUACAGCCAUGGUCGCCCCGCAGCACGCCCACGCUC
          Chromosome Location Chr. 10 - 62014995 - 62015095 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0017123
          miRBase ID: rno-miR-132-5p
          Mature miRNA Sequence: ACCGUGGCUUUCGAUUGUUACU
          Chromosome Location: Chr. 10 - 62014995 - 62015095 [+] on Build Rnor_6.0

          Cricetulus griseus

          Stem-loop ID cgr-mir-132
          Stem-loop Accession # MI0020396
          Stem-loop Sequence
          UCCAGGGCAACCGUGGCUUUCGAUUGUUACUGUGGGAACCGGAGGUAACAGUCUACAGCCAUGGUCGCCCCGCAGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023758
          miRBase ID: cgr-miR-132-5p
          Mature miRNA Sequence: ACCGUGGCUUUCGAUUGUUACU
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-132
          Stem-loop Accession # MI0038976
          Stem-loop Sequence
          ACCGUGGCUUUCGAUUGUUACUGUGGAAACCGGAGGUAACAGUCUACAGCCAUGGUCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047631
          miRBase ID: dno-miR-132-5p
          Mature miRNA Sequence: ACCGUGGCUUUCGAUUGUUACU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-132
          Stem-loop Accession # MI0038635
          Stem-loop Sequence
          ACCGUGGCUUUCGAUUGUUACUGUGGGAACCGGAGGUAACAGUCUACAGCCAUGGUCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046984
          miRBase ID: cpo-miR-132-5p
          Mature miRNA Sequence: ACCGUGGCUUUCGAUUGUUACU
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-132
          Stem-loop Accession # MI0005338
          Stem-loop Sequence
          GGGCAACCGUGGCUUUCGAUUGUUACUGUGGGAACCAGGGGUAACAGUCUACAGCCAUGGUCGCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026693
          miRBase ID: mdo-miR-132-5p
          Mature miRNA Sequence: ACCGUGGCUUUCGAUUGUUACU
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-132
          Stem-loop Accession # MI0032437
          Stem-loop Sequence
          ACCGUGGCUUUCGAUUGUUACUGUGGGAACCGGAGGUAACAGUCUACAGCCAUGGUCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039991
          miRBase ID: pal-miR-132-5p
          Mature miRNA Sequence: ACCGUGGCUUUCGAUUGUUACU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-132
          Stem-loop Accession # MI0007620
          Stem-loop Sequence
          CCGCCCCCGCGUCUCCAGGGCAACCGUGGCUUUCGAUUGUUACUGUGGGAACUGGAGGUAACAGUCUACAGCCAUGGUCGCCCCGCAGCACGCCCACGCGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026816
          miRBase ID: mml-miR-132-5p
          Mature miRNA Sequence: ACCGUGGCUUUCGAUUGUUACU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303111_pri
          TaqMan™ Pri-miRNA Assay : Rn03465057_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12827
          Ambion® Pre-miR™ miRNA Precursor : PM12827
          TaqMan™ MicroRNA Assay : 002132
          mirVana® miRNA inhibitor : MH12827
          mirVana® miRNA mimic : MC12827
          TaqMan™ Advanced miRNA Assay : rno481320_mir
          FGMIR : TaqManTM Advanced miRNA Human B Card
          FGMIR : QS TaqmanTM OpenArray Human Advanced miRNA Panel
          FGMIR : TaqManTM Advanced miRNA Human B 96-well Plate 2
          MIRNA : TaqMan® Advanced miRNA Human B 96-well Plates, Standard
          MIRNA : TaqMan® Advanced miRNA Human B Card
          MIRNA : TaqMan® Advanced miRNA Human A and B 96-well Plates, Standard
          MIRNA : TaqMan® OpenArray® Human Advanced MicroRNA Panel
          FGMIR : TaqManTM Advanced miRNA Human B 96-well Plate 2
          MIRNA : TaqMan® Advanced miRNA Human A and B Cards

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline