Search
Search

| Assay Name: | hsa-miR-134-3p |
|---|---|
| Stem-loop Accession Number: | MI0000474 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CCUGUGGGCCACCUAGUCACCAA |
| Species: | Human, Rhesus monkey |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000112, mir-134 |
Mature miRNA Sequence: | CCUGUGGGCCACCUAGUCACCAA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-134-3p | MIMAT0026481 |
| Rhesus monkey | mml-miR-134-3p | MIMAT0026819 |