Search
Search

| Assay Name: | hsa-miR-520g-3p |
|---|---|
| Stem-loop Accession Number: | MI0003166 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | ACAAAGUGCUUCCCUUUAGAGUGU |
| Species: | Human, Gorilla gorilla, Pan troglodytes, Pongo pygmaeus |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000020, mir-515 |
Mature miRNA Sequence: | ACAAAGUGCUUCCCUUUAGAGUGU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-520g-3p | MIMAT0002858 |
| Gorilla gorilla | ggo-miR-520g | MIMAT0036434 |
| Pan troglodytes | ptr-miR-520g | MIMAT0008203 |
| Pan troglodytes | ptr-miR-520h | MIMAT0008204 |
| Pongo pygmaeus | ppy-miR-520g | MIMAT0015975 |