Search
Search

| Assay Name: | hsa-miR-4429 |
|---|---|
| Stem-loop Accession Number: | MI0016768 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AAAAGCUGGGCUGAGAGGCG |
| Species: | Human |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0001474, mir-4429 |
Mature miRNA Sequence: | AAAAGCUGGGCUGAGAGGCG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-4429 | MIMAT0018944 |